Skip to Content
Merck
All Photos(1)

Key Documents

30-4890

Sigma-Aldrich

2,3,5-Triphenyltetrazolium chloride

JIS special grade, 98.0-102.0%

Synonym(s):

TPTZ, TTZ, Tetrazolium Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15ClN4
CAS Number:
Molecular Weight:
334.80
Beilstein:
3923356
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
grade:
JIS special grade
application(s):
microbiology
Pricing and availability is not currently available.

grade

JIS special grade

Assay

98.0-102.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

application(s)

microbiology

SMILES string

[Cl-].c1ccc(cc1)-c2nn(-c3ccccc3)[n+](n2)-c4ccccc4

InChI

1S/C19H15N4.ClH/c1-4-10-16(11-5-1)19-20-22(17-12-6-2-7-13-17)23(21-19)18-14-8-3-9-15-18;/h1-15H;1H/q+1;/p-1

InChI key

PKDBCJSWQUOKDO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Zhang et al.
Neurochemical research, 39(7), 1363-1373 (2014-05-06)
Phosphorylated tau was found to be regulated after cerebral ischemia and linked to high risk for the development of post-stroke dementia. Our previous study showed that ginsenoside Rd (Rd), one of the main active ingredients in Panax ginseng, decreased tau
Xufeng Tao et al.
Free radical biology & medicine, 84, 103-115 (2015-03-17)
We previously reported the promising effect of dioscin against hepatic ischemia/reperfusion (I/R) injury, but its effect on cerebral I/R injury remains unknown. In this work, an in vitro oxygen-glucose deprivation and reoxygenation (OGD/R) model and an in vivo middle cerebral
Liyan Ping et al.
PloS one, 7(6), e38427-e38427 (2012-06-08)
Tetrazolium salts are widely used in biology as indicators of metabolic activity - hence termed vital dyes - but their reduction site is still debated despite decades of intensive research. The prototype, 2,3,5- triphenyl tetrazolium chloride, which was first synthesized
Joana Arbeloa et al.
Neurobiology of disease, 45(3), 954-961 (2011-12-22)
Overactivation of subtype P2X7 receptors can induce excitotoxic neuronal death by calcium (Ca(2+)) overload. In this study, we characterize the functional properties of P2X7 receptors using electrophysiology and Ca(2+) monitoring in primary cortical neuron cultures and in brain slices. Both
Y Yang et al.
NMR in biomedicine, 25(1), 177-188 (2011-10-01)
The aim of the study was the testing of sustained intrapericardial delivery of vascular growth factors (GFs) from alginate beads on cryoinjury size and perfusion. In domestic pigs (15-20 kg, n = 21), the left ventricular (LV) anterolateral wall of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service