Skip to Content
Merck
All Photos(1)

Key Documents

30-3670

Sigma-Aldrich

D-(+)-Trehalose dihydrate

SAJ special grade, ≥99.0%

Synonym(s):

α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
5322018
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Explanation and prediction for the product of trehalose dihydrate selective dehydration process using mid-frequency Raman difference spectra.: This research employs mid-frequency Raman difference spectra to analyze and predict the products formed during the selective dehydration of D-(+)-Trehalose dihydrate, providing insights into its structural changes during the dehydration process (Fan et al., 2024).

  • Physico-geometrical Kinetic Aspects of the Thermal Dehydration of Trehalose Dihydrate.: This study investigates the kinetic and geometrical aspects of the thermal dehydration of D-(+)-Trehalose dihydrate, offering detailed understanding of its stability and transformation under thermal conditions (Hara et al., 2022).

  • Trehalose and its dihydrate: terahertz insights from solid to solution states.: Utilizing terahertz time-domain spectroscopy, this paper explores the spectral characteristics of D-(+)-Trehalose dihydrate in both solid and solution states, comparing its behaviors and providing a comprehensive analysis through density functional theory simulations (Zhang et al., 2024).

  • Oiling out and Solubility of Trehalose Dihydrate in Water + Isopropanol/1-Propanol/tert-Butanol Solvents from 283.15 to 328.15 K.: This research examines the solubility and oiling out behavior of D-(+)-Trehalose dihydrate in various solvent mixtures over a range of temperatures, providing valuable data for its application in different solvent systems (Yu et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Ullrich et al.
Journal of pharmaceutical sciences, 104(6), 2040-2046 (2015-04-03)
The importance of cake adhesion to the inside vial wall during lyophilization of amorphous trehalose cakes was determined by using hydrophobized vials. The degrees of cake shrinkage and cracking were determined independently by photographic imaging of the cake top surface
Alan Twomey et al.
International journal of pharmaceutics, 487(1-2), 91-100 (2015-04-19)
In frozen and lyophilized systems, the biological to be stabilized (e.g. therapeutic protein, biomarker, drug-delivery vesicle) and the cryo-/lyo-protectant should be co-localized for successful stabilization. During freezing and drying, many factors cause physical separation of the biological from the cryo-/lyo-protectant
Sabine Ullrich et al.
Pharmaceutical research, 32(8), 2503-2515 (2015-02-06)
Measurement of the kinetic development of shrinkage and cracking of an amorphous trehalose cake as they take place during lyophilization. A novel technique has been developed which monitors a vial in situ during the freeze-drying cycle. The 2-dimensional degrees of
Matthias Erber et al.
International journal of pharmaceutics, 495(2), 692-700 (2015-09-24)
A cryopellet formulation of the diagnostic protein ecarin has been developed that is suitable for use to monitor blood coagulation via in vitro thromboelastometry. The coagulation activity of the ecarin was measured by thromelastometry using whole blood. In aqueous solution
Sabine Ullrich et al.
Journal of pharmaceutical sciences, 104(1), 155-164 (2014-11-26)
An experimental technique is presented to determine independently shrinkage and cracking in lyophilized amorphous cakes based on photographic imaging of their top surface. An inverse correlation between cake shrinkage and cracking during freeze-drying is seen. Shrinkage relaxes the drying tension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service