Reactions of the allyltin trichloride 45 generated from (4S)-4-benzyloxypent-2-enyl(tributyl)stannane 1 with imines prepared from glyoxylates proceed with useful levels of 1,5-stereocontrol in favour of (4E)-2,6-anti-2-(alkylamino)-6-benzyloxyhept-4-enoates 49. This stereoselectivity, controlled by the chirality of the stannane, dominates over any intrinsic stereochemical
A six-step one-pot reaction was designed for synthesizing homodimeric 7-phenylindolo[3,2-a]carbazoles from 1H-indoles and β-nitrostyrenes, in the presence of SnCl(2)·2H(2)O. The reactions proceeded under very mild conditions and the desired heterocycles were obtained in moderate to good yields. An unprecedented mechanism
Journal of the American Chemical Society, 134(10), 4998-5003 (2012-02-15)
Cationic and anionic species of heavier low-valent group 14 elements are intriguing targets in main group chemistry due to their synthetic potential and industrial applications. In the present study, we describe the synthesis of cationic (MCl(+)) and anionic (MCl(3)(-)) species
International journal of nanomedicine, 6, 3077-3085 (2011-12-14)
The purpose of this study was to develop intraperitoneal hyperthermic therapy based on magnetic fluid hyperthermia, nanoparticle-wrapped cisplatin chemotherapy, and magnetic particles of albumin. In addition, to combine the multiple-killing effects of hyperthermal targeting therapy, chemotherapy, and radiotherapy, the albumin-nanoparticle
The anionic speciation of chlorostannate(II) ionic liquids, prepared by mixing 1-alkyl-3-methylimidazolium chloride and tin(II) chloride in various molar ratios, χ(SnCl2), was investigated in both solid and liquid states. The room temperature ionic liquids were investigated by (119)Sn NMR spectroscopy, X-ray