Skip to Content
Merck
All Photos(1)

Key Documents

28-4340

Sigma-Aldrich

Sodium sulfite

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥95.0%

form

solid

availability

available only in Japan

pH

9.0-10.5 (25 °C, 126 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Krickl et al.
Physical chemistry chemical physics : PCCP, 19(35), 23773-23780 (2017-07-01)
In this contribution, we (i) link the mesoscopic structuring of the binary structured solvent mixture H
Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Jingang Wang et al.
Chemosphere, 91(3), 351-357 (2012-12-26)
This paper presents a simple, easy and reliable method for determination of EDTA (ethylenediaminetetraacetic acid) in aqueous system. Using EDTA chelating with ferric irons, an excessive amount of Fe(3+) was added to EDTA solution, and Fe(3+) in excess was reduced
Fabian Leinisch et al.
Chemical research in toxicology, 24(12), 2217-2226 (2011-10-19)
Nitrone spin traps such as 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) are commonly used for free radical detection. Though proven examples are rare, artifact formation must be considered. For example, the Forrester-Hepburn mechanism yields the same radical adduct as that formed by genuine radical
Christian Ciardi et al.
The British journal of nutrition, 107(6), 826-833 (2011-08-02)
Obesity leads to the activation of pro-inflammatory pathways, resulting in a state of low-grade inflammation. Recently, several studies have shown that the exposure to lipopolysaccharide (LPS) could initiate and maintain a chronic state of low-grade inflammation in obese people. As

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service