All Photos(1)
About This Item
Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
liquid
Recommended Products
form
liquid
availability
available only in Japan
concentration
0.05 M
1/10 N
storage temp.
15-25°C
SMILES string
[Na+].[Na+].[O-]C(=O)C([O-])=O
InChI
1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2
InChI key
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Juntaek Oh et al.
The Journal of biological chemistry, 289(16), 11465-11475 (2014-03-13)
The Burkholderia species utilize acetyl-CoA and oxaloacetate, substrates for citrate synthase in the TCA cycle, to produce oxalic acid in response to bacterial cell to cell communication, called quorum sensing. Quorum sensing-mediated oxalogenesis via a sequential reaction by ObcA and
Lu Gao et al.
International journal of phytoremediation, 15(4), 307-319 (2013-03-16)
Phytolacca americana L. can accumulate large amounts of heavy metals in its aerial tissues, especially cadmium (Cd) and manganese (Mn). It has great potential for use in phytoextraction of metals from multi-metal-contaminated soils. This study was conducted to further investigate
Taiying Zhang et al.
Carbohydrate polymers, 92(1), 334-344 (2012-12-12)
Dilute oxalic acid pretreatment was applied to maple wood to improve compatibility with downstream operations, and its performance in pretreatment and subsequent enzymatic hydrolysis was compared to results for hydrothermal and dilute hydrochloric and sulfuric acid pretreatments. The highest total
Lois Fernandez et al.
Clinical biochemistry, 46(4-5), 285-288 (2012-12-12)
There are no direct comparisons of blood glucose values in samples collected with barrier serum tubes (SST) and NaF/potassium oxalate (NaF/KOx) plasma tubes. Collection of samples in SST tubes can offer considerable savings and specimen processing advantages for national level
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service