Recommended Products
availability
available only in Japan
dilution
(for analytical testing)
Suitability
for electrophoresis
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pushpa Patel et al.
Reproductive biology and endocrinology : RB&E, 7, 10-10 (2009-02-10)
Hydrogen sulphide is a gas signalling molecule which is produced endogenously from L-cysteine via the enzymes cystathionine beta-synthase (CBS) and cystathionine gamma-lyase (CSE). The possible role of hydrogen sulphide in reproduction has not yet been fully investigated. It has been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service