Skip to Content
Merck
All Photos(1)

Key Documents

24-3300

Sigma-Aldrich

Piperidine

SAJ first grade, ≥99.0%

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein:
102438
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
SAJ first grade
bp:
106 °C (lit.)
vapor pressure:
23 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.452 (lit.)

bp

106 °C (lit.)

mp

−13 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Łuczkowski et al.
Inorganic chemistry, 47(23), 10875-10888 (2008-10-31)
A de novo protein design strategy provides a powerful tool to elucidate how heavy metals interact with proteins.Cysteine derivatives of the TRI peptide family (Ac-G(LKALEEK)4G-NH2) have been shown to bind heavy metals in an unusual trigonal geometry. Our present objective
Total synthesis of (-)-kopsinine by an asymmetric one-pot [n+2+3] cyclization.
Shingo Harada et al.
Chemistry, an Asian journal, 7(10), 2196-2198 (2012-08-22)
J Phillip Turner et al.
ACS chemical neuroscience, 5(7), 552-558 (2014-04-03)
Alzheimer's disease (AD) is the most common form of dementia and the sixth leading cause of death in the United States. Plaques composed of aggregated amyloid-beta protein (Aβ) accumulate between the neural cells in the brain and are associated with
Xuwang Chen et al.
Bioorganic & medicinal chemistry, 20(12), 3856-3864 (2012-05-18)
A novel series of piperidine-linked amino-triazine derivatives were designed, synthesized and evaluated for in vitro anti-HIV activity as non-nucleoside reverse transcriptase inhibitors on the basis of our previous work. Screening results indicated that most compounds showed excellent activity against wild-type
Shruti U Gandhy et al.
BMC cancer, 12, 564-564 (2012-12-01)
Curcumin inhibits growth of several cancer cell lines, and studies in this laboratory in bladder and pancreatic cancer cells show that curcumin downregulates specificity protein (Sp) transcription factors Sp1, Sp3 and Sp4 and pro-oncogenic Sp-regulated genes. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service