Skip to Content
Merck
All Photos(1)

Key Documents

24-2760

Sigma-Aldrich

Phosphorus pentoxide

JIS special grade, ≥98.0%

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
MDL number:
UNSPSC Code:
12352303

grade

JIS special grade

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

Assay

≥98.0%

form

crystalline

availability

available only in Japan

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrício Aulo Ogliari et al.
Journal of dentistry, 36(3), 171-177 (2008-02-05)
The aim of this study was to synthesize an acidic monomer using an alternative synthetic pathway and to evaluate the influence of the acidic monomer concentration on the microtensile bond strength to dentin. The intermediary 5-hydroxypentyl methacrylate (HPMA) was synthesized
Ana García et al.
Acta biomaterialia, 7(3), 1265-1273 (2010-10-19)
Herein we report for the first time the synthesis of three-dimensional scaffolds in the binary system SiO2-P2O5 exhibiting different scales of porosity: (i) highly ordered mesopores with diameters of ca. 4 nm; (ii) macropores with diameters in the 30-80 μm
J Ma et al.
Colloids and surfaces. B, Biointerfaces, 81(1), 87-95 (2010-07-30)
Mesoporous 58SiO(2)-(38-x)CaO-xMgO-4P(2)O(5) glasses (where x=0, 5, 10 and 20 mol%) have been prepared by the sol-gel synthesis route. The effects of the substitution of MgO for CaO on glass degradation and bioactivity were studied in tris-(hydroxymethyl)-aminomethane and hydrochloric acid buffer
Ping-Jian Li et al.
Nano letters, 9(7), 2513-2518 (2009-07-09)
The single-crystal n-type and p-type ZnO nanowires (NWs) were synthesized via a chemical vapor deposition method, where phosphorus pentoxide was used as the dopant source. The electrical and photoluminescence studies reveal that phosphorus-doped ZnO NWs (ZnO:P NWs) can be changed
Devalina Ray et al.
Carbohydrate research, 346(14), 2145-2148 (2011-09-06)
A convenient, mild and environment-friendly dehydration reaction of fructose in ionic liquid using phosphorous pentoxide (P(2)O(5)) has been investigated. The acidic nature of P(2)O(5) along with its hygroscopic properties has been successfully utilized to afford 81.2% yield of 5-hydroxymethylfurfural (5-HMF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service