Skip to Content
Merck
All Photos(1)

Key Documents

24-1980

Sigma-Aldrich

Phenylhydrazine hydrochloride

JIS special grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNH2 · HCl
CAS Number:
Molecular Weight:
144.60
Beilstein:
3594958
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

JIS special grade

Assay

≥98.5%

availability

available only in Japan

mp

250-254 °C (dec.) (lit.)

SMILES string

Cl[H].NNc1ccccc1

InChI

1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H

InChI key

JOVOSQBPPZZESK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Frazer et al.
British journal of haematology, 157(5), 615-626 (2012-03-28)
The BMP/SMAD signalling pathway plays an important role in iron homeostasis, regulating hepcidin expression in response to body iron levels. However, the role of this pathway in the reduction in hepcidin associated with increased erythropoiesis (and secondary iron loading) is
Jeanne E Hendrickson et al.
Transfusion, 51(11), 2445-2454 (2011-05-17)
Fourteen-day stored red blood cells (RBCs) containing an RBC-specific transgenic antigen (HOD) induce a recipient proinflammatory cytokine storm and are significantly more immunogenic compared to fresh RBCs. Given that recipient mice clear transfused stored RBCs more rapidly than fresh RBCs
Yuji Sonoda et al.
Cells, tissues, organs, 196(6), 555-564 (2012-06-29)
Fetal liver hematopoiesis in mice disappears approximately 2 weeks after birth; however, under experimental acute anemia extramedullary hematopoiesis occurs in the livers of adult mice. The hematopoietic foci in the liver during extramedullary hematopoiesis contain erythroblasts and macrophages. In this
Snežana D Marković et al.
Acta biologica Hungarica, 62(2), 122-132 (2011-05-11)
Stimulated erythropoiesis and reticulocytosis can be induced by daily bleeding, or by phenylhydrazine (PHZ) treatment. We compared the in vivo effects of PHZ and bleeding treatment on haematological, energy and redox status parameters in red blood cells (RBC) of rats.
Claudia Köthe et al.
Inorganic chemistry, 51(18), 9740-9747 (2012-09-07)
The reaction of [L(tBu)Ni(OEt(2))] (where L(tBu) = [HC(C(t)BuNC(6)H(3)((i)Pr)(2))(2)](-)) with phenylhydrazine leads to the phenylhydrazido(1-) complex [L(tBu)Ni(η(2)-NPhNH(2))] (1) with concomitant formation of H(2). Treatment of 1 with potassium graphite in the presence of crown ether again leads to H(2) evolution and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service