Skip to Content
Merck
All Photos(1)

Key Documents

24-1020

Sigma-Aldrich

Perchloric acid

JIS special grade

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

form

liquid

reaction suitability

reagent type: oxidant

availability

available only in Japan

concentration

70.0-72.0%

density

167 g/cm3

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Greenwood et al.
ACS nano, 9(5), 5520-5535 (2015-04-22)
We shine light on the covalent modification of graphite and graphene substrates using diazonium chemistry under ambient conditions. We report on the nature of the chemical modification of these graphitic substrates, the relation between molecular structure and film morphology, and
Martin G Liebensteiner et al.
Science (New York, N.Y.), 340(6128), 85-87 (2013-04-06)
Perchlorate and chlorate anions [(per)chlorate] exist in the environment from natural and anthropogenic sources, where they can serve as electron acceptors for bacteria. We performed growth experiments combined with genomic and proteomic analyses of the hyperthermophile Archaeoglobus fulgidus that show
Dipanwita Das et al.
Journal of the American Chemical Society, 135(10), 4018-4026 (2013-02-28)
Catalytic four-electron reduction of O2 by ferrocene (Fc) and 1,1'-dimethylferrocene (Me2Fc) occurs efficiently with a dinuclear copper(II) complex [Cu(II)2(XYLO)(OH)](2+) (1), where XYLO is a m-xylene-linked bis[(2-(2-pyridyl)ethyl)amine] dinucleating ligand with copper-bridging phenolate moiety], in the presence of perchloric acid (HClO4) in
Dipanwita Das et al.
Journal of the American Chemical Society, 135(7), 2825-2834 (2013-02-12)
Selective two-electron plus two-proton (2e(-)/2H(+)) reduction of O(2) to hydrogen peroxide by ferrocene (Fc) or 1,1'-dimethylferrocene (Me(2)Fc) in the presence of perchloric acid is catalyzed efficiently by a mononuclear copper(II) complex, [Cu(II)(tepa)](2+) (1; tepa = tris[2-(2-pyridyl)ethyl]amine) in acetone. The E(1/2)
Kristiina Cajanus et al.
The journal of pain : official journal of the American Pain Society, 15(12), 1248-1256 (2014-09-23)
Most clinically used opioids are mu-opioid receptor agonists. Therefore, genetic variation of the OPRM1 gene that encodes the mu-opioid receptor is of great interest for understanding pain management. A polymorphism 118A>G (rs1799971) within the OPRM1 gene results in a missense

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service