Skip to Content
Merck
All Photos(1)

Key Documents

23-0320

Sigma-Aldrich

Corn oil

CP

Synonym(s):

Maize oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

grade

CP

form

liquid

availability

available only in Japan

functional group

ester

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

489.2 °F

Flash Point(C)

254 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa D Yee et al.
Molecular nutrition & food research, 57(2), 320-327 (2012-12-06)
We previously demonstrated that lifelong feeding of diets enriched in n-3 fatty acids such as docosahexaenoic acid (DHA) and eicosapentaenoic acid (EPA) significantly inhibits HER-2/neu-mediated mammary tumorigenesis in mice. Of interest is whether dietary n-3 fatty acids exert effects at
B G Kim et al.
Journal of animal science, 91(2), 755-763 (2012-11-14)
An experiment was conducted to determine the true ileal digestibility (TID) and the true total tract digestibility (TTTD) of acid-hydrolyzed ether extract (AEE) in extracted corn oil, high-oil corn, distillers dried grains with solubles (DDGS), corn germ, and high protein
Hui Li et al.
Nature genetics, 45(1), 43-50 (2012-12-18)
Maize kernel oil is a valuable source of nutrition. Here we extensively examine the genetic architecture of maize oil biosynthesis in a genome-wide association study using 1.03 million SNPs characterized in 368 maize inbred lines, including 'high-oil' lines. We identified
Quanri Jin et al.
Carcinogenesis, 29(8), 1614-1622 (2008-07-19)
The inhibitor of apoptosis protein survivin is selectively expressed in tumor cells. The tobacco component nicotine increases the transcription of the survivin gene in non-small cell lung cancer cells. However, the role of survivin expression induced by tobacco component is
Hua-Min Liu et al.
Bioresource technology, 128, 58-64 (2012-12-01)
Hydrothermal liquefaction of cornstalk at 180-300 °C at ratios of water to cornstalk of 6-14 was conducted, and the reaction products were lumped into gas, water-soluble organics (ethanol-insoluble and ethanol-soluble organics), heavy oil, volatile organic compounds, and acid-soluble and acid-insoluble

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service