Skip to Content
Merck
All Photos(1)

Key Documents

19-4490

Sigma-Aldrich

Dimethyl yellow

JIS special grade

Synonym(s):

4-(Dimethylamino)azobenzene, N,N-Dimethyl-4-(phenylazo)aniline, Butter yellow, Methyl yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4N(CH3)2
CAS Number:
Molecular Weight:
225.29
Colour Index Number:
11020
Beilstein:
746016
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
grade:
JIS special grade
Pricing and availability is not currently available.

grade

JIS special grade

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

111 °C (dec.) (lit.)

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccc2

InChI

1S/C14H15N3/c1-17(2)14-10-8-13(9-11-14)16-15-12-6-4-3-5-7-12/h3-11H,1-2H3

InChI key

JCYPECIVGRXBMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debadutta Mishra et al.
Toxicology letters, 203(1), 40-47 (2011-03-08)
The azo-dye p-dimethylaminoazobenzene (p-DAB) is a potential tumor initiator in rodents, but the underlying mechanism is not clear. Following chronic feeding of the carcinogen for 3, 5 and 7 weeks, trace elemental status, free radical generation, oxidative damage, antioxidant profile
Michalina Oplatowska et al.
The Analyst, 136(11), 2403-2410 (2011-04-22)
Sensitive and specific enzyme-linked immunosorbent assays (ELISAs) were developed for the detection of two illegal synthetic dyes: Methyl Yellow (MY) and Rhodamine B (RB) in food. Polyclonal antibodies were raised against synthesised immunogens and employed in unique direct disequilibrium ELISAs.
Akira Abe et al.
Analytical biochemistry, 434(1), 78-83 (2012-11-14)
Lysosomal phospholipase A2 (group XV PLA2, LPLA2) is a lysosomal enzyme linked to drug-induced phospholipidosis. We developed phospholipid "smart probes" based on the conversion of a quenched fluorogenic substrate to a fluorescent product. Due to the preference of LPLA2 for
Derek Ho et al.
Biochemistry, 50(22), 4830-4842 (2011-05-03)
The membrane topology of the colicin E1 channel domain was studied by fluorescence resonance energy transfer (FRET). The FRET involved a genetically encoded fluorescent amino acid (coumarin) as the donor and a selectively labeled cysteine residue tethered with DABMI (4-(dimethylamino)phenylazophenyl-4'-maleimide)
Ethiraju Babu et al.
Journal of fluorescence, 23(1), 137-146 (2012-09-12)
We describe here the construction of a sensitive and selective optical sensor system for the detection of human α-thrombin. The surface functionalized luminescent [Ru(dpsphen)(3)](4-) (dpsphen-4,7-diphenyl-1,10-phenanthroline disulfonate) ion doped silica nanoparticles (SiNPs) with a size ~70 nm have been prepared. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service