Environmental science and pollution research international, 19(9), 3696-3708 (2012-10-12)
TiO(2):Nb nanopowders within a dopant concentration in the range of 0.1-15 at.% were prepared by one-step flame spray synthesis. Effect of niobium doping on structural, optical and photocatalytic properties of titanium dioxide nanopowders was studied. Morphology and structure were investigated by
Journal of hazardous materials, 239-240, 316-324 (2012-09-29)
Novel p-n junction photocatalysts BiOI/(BiO)2CO3 with different contents of BiOI were in situ synthesized by etching (BiO)2CO3 precursor with hydroiodic acid (HI) solution. X-ray diffraction (XRD), field emission scanning electron microscopy (FE-SEM), Fourier transform infrared spectrometry (FT-IR), energy-dispersive spectroscopy (EDS)
Environmental science and pollution research international, 19(9), 3743-3750 (2012-10-12)
Solar wastewater treatment based on photocatalytic reactions is a green process that utilizes renewable energy resources and minimizes secondary pollution. Reactor design plays an important role in promoting treatment efficiency and throughput density (based on unit volume of the reactor).
The structure, composition and photocatalytic activity of TiO(2) nanoparticles annealed in various gas atmospheres (N(2), NH(3) and H(2)) were studied in this work. The effect of treatment on crystal structure, particle size, chemical composition and optical absorbance were assessed by
Colloids and surfaces. B, Biointerfaces, 103, 658-661 (2012-12-26)
In this paper, we report on biosynthesis of silver nanoparticles using Ulva lactuca (seaweed) at room temperature along with photocatalytic degradation of methyl orange dye. UV spectral analysis showed peak at 430 nm with special reference to the excitation of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.