Skip to Content
Merck
All Photos(1)

Documents

19-3180

Sigma-Aldrich

N,N′-Methylenebis(acrylamide)

≥99.0%

Synonym(s):

Bis-acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
41105305
PubChem Substance ID:

vapor density

5.31 (vs air)

Assay

≥99.0%

form

solid

availability

available only in Japan

technique(s)

electrophoresis: suitable

mp

>300 °C (lit.)

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for electrophoresis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Aho et al.
British journal of anaesthesia, 115(2), 258-266 (2015-07-04)
Concomitantly recorded Bispectral Index® (BIS) and Entropy™ values sometimes show discordant trends during general anaesthesia. Previously, no attempt had been made to discover which EEG characteristics cause discrepancies between BIS and Entropy. We compared BIS and Entropy values, and analysed
Jia Liu et al.
Carbohydrate polymers, 94(1), 539-546 (2013-04-03)
To better use wheat straw and minimize its negative impact on environment, a novel semi-interpenetrating polymer networks (semi-IPNs) superabsorbent resin (SAR) composed of wheat straw cellulose-g-poly (potassium acrylate) (WSC-g-PKA) network and linear polyvinyl alcohol (PVA) was prepared by polymerization in
Han Gao et al.
Macromolecular rapid communications, 34(1), 63-68 (2012-10-20)
Traditional pH-sensitive hydrogels inevitably suffer strength deterioration while the responsive weak acid or base groups are in the ionized state. In this study, we report on a facile approach to fabricate a novel pH-sensitive high-strength hydrogel from copolymerization of two
David R Kryscio et al.
Analytica chimica acta, 718, 109-115 (2012-02-07)
Molecularly imprinted polymers are synthetic antibody mimics formed by the crosslinking of organic or inorganic polymers in the presence of an analyte which yields recognitive polymer networks with specific binding pockets for that biomolecule. Surface imprinted polymers were synthesized via
Jean Guglielminotti et al.
Anesthesiology, 122(5), 985-993 (2015-03-03)
Individual assessment of the amplitude of a physiologic reflex evoked by a standardized noxious test (SNT) before surgical stimulation has been suggested to predict movement upon the forthcoming surgical stimulation. This study aimed to compare the ability of pupillary dilatation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service