Skip to Content
Merck
All Photos(1)

Key Documents

19-1880

Sigma-Aldrich

Mercury(II) sulfate

JIS special grade, ≥99.0%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
crystalline
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥99.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: mercury

availability

available only in Japan

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shovana Pradhan et al.
Chemosphere, 136, 198-203 (2015-05-24)
Reverse osmosis (RO) concentrate (ROC) streams generated from RO-based municipal wastewater reclamation processes pose potential health and environmental risks on their disposal to confined water bodies such as bays. A UV/H2O2 advanced oxidation process followed by a biological activated carbon
Paul I Dargan et al.
Critical care (London, England), 7(3), R1-R6 (2003-06-10)
Inorganic mercury poisoning is uncommon, but when it occurs it can result in severe, life-threatening features and acute renal failure. Previous reports on the use of extracorporeal procedures such as haemodialysis and haemoperfusion have shown no significant removal of mercury.
Necati Kayaalp et al.
Environmental science and pollution research international, 17(9), 1547-1552 (2010-05-25)
Chemical oxygen demand (COD) is used as a discharge standard parameter in wastewater treatment plant design, environmental modelling and many other applications. Chloride interference is an important problem of COD measurement for wastewaters containing low organic matter and high chloride
L A Ivanova et al.
Gigiena i sanitariia, (11)(11), 66-67 (1991-11-01)
In experiments with two tissue culture lines it was shown, that cytotoxicity of mercury (1) derivatives (measured by CL50) is lesser two times, than it was for mercury (2) derivates. No significant differences in cytotoxicity were noted by the cell
A P de Souza et al.
Dental materials : official publication of the Academy of Dental Materials, 16(2), 103-108 (2001-02-24)
The interaction between metal ions and the oral environment is a major subject matter in dental research. Matrix metalloproteinases (MMPs) have been implicated in several pathologic oral processes such as periodontal tissue destruction, root caries, tumour invasion and temporomandibular joint

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service