Skip to Content
Merck
All Photos(1)

Key Documents

19-1540

Sigma-Aldrich

Mercury(II) acetate

JIS special grade, ≥99.0%

Synonym(s):

Mercuric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Hg
CAS Number:
Molecular Weight:
318.68
Beilstein:
3563831
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
powder

grade

JIS special grade

Assay

≥99.0%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

availability

available only in Japan

mp

179-182 °C (lit.)

SMILES string

CC(=O)O[Hg]OC(C)=O

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris A Trofimov et al.
Dalton transactions (Cambridge, England : 2003), 41(18), 5512-5516 (2012-03-21)
N-Vinylpyrroles and -indoles bearing electron-withdrawing substituents at the pyrrole ring are mercurated, with 1 equivalent of Hg(OAc)(2) in dry MeCN (20-80 °C), regioselectively at the vinyl group (yields are almost quantitative), while their congeners without electron-withdrawing functions are mercurated both
Y Hojima et al.
Matrix biology : journal of the International Society for Matrix Biology, 14(2), 113-120 (1994-03-01)
Procollagen C- and N-proteinases specifically cleave the C- and N-terminal extension propeptides of type I, II and III procollagen molecules. The collagen molecules generated by the enzymes self-assemble into collagen fibrils. We previously observed the inhibition of these enzymes purified
H O Yang et al.
Glycobiology, 10(10), 1033-1039 (2000-10-13)
Eight oligosaccharides were prepared from dermatan sulfate (DS) and their structures were elucidated. Porcine intestinal mucosal DS was subjected to controlled depolymerization using chondroitin ABC lyase (chondroitinase ABC). The oligosaccharide mixture formed was fractionated by low-pressure gel permeation chromatography (GPC).
An efficient synthesis of 3,4,6-tri-O-benzyl-2-C-methyl-D-glucal.
R W Scott et al.
Carbohydrate research, 291, 205-208 (1996-09-23)
Yukio Hitotsuyanagi et al.
The Journal of organic chemistry, 67(10), 3266-3271 (2002-05-11)
A novel cis amide bond surrogate incorporating 1,2,4-triazole was designed and synthesized by the reaction of a thionotripeptide, formic hydrazide, and mercury(II) acetate. This method of surrogate formation was also applicable to a cyclic thionopeptide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service