Skip to Content
Merck
All Photos(1)

Key Documents

19-1330

Sigma-Aldrich

2-Mercaptoethanol

SAJ special grade, ≥99.0%

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein:
773648
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
grade:
SAJ special grade
bp:
157 °C (lit.)
vapor pressure:
1 mmHg ( 20 °C)

grade

SAJ special grade

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

expl. lim.

18 %

availability

available only in Japan

concentration

14.3 M (pure liquid)

dilution

(for analytical testing)

refractive index

n20/D 1.500 (lit.)

pH

4.5-6 (20 °C, 500 g/L)

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Rothe et al.
Blood, 123(23), 3622-3634 (2014-04-24)
Previous studies demonstrated that imatinib mesylate (IM) induces autophagy in chronic myeloid leukemia (CML) and that this process is critical to cell survival upon therapy. However, it is not known if the autophagic process differs at basal levels between CML
Leon Tribolet et al.
The Journal of infectious diseases, 211(3), 416-425 (2014-08-21)
Na-ASP-2 is an efficacious hookworm vaccine antigen. However, despite elucidation of its crystal structure and studies addressing its immunobiology, the function of Na-ASP-2 has remained elusive. We probed a 9000-protein human proteome microarray with Na-ASP-2 and showed binding to CD79A
E M Michalak et al.
Cell death and differentiation, 15(6), 1019-1029 (2008-02-09)
The ability of p53 to induce apoptosis in cells with damaged DNA is thought to contribute greatly to its tumour suppressor function. P53 has been proposed to induce apoptosis via numerous transcriptional targets or even by direct cytoplasmic action. Two
D Zhu et al.
Leukemia, 29(7), 1587-1599 (2015-02-14)
The pathogenesis of Chlamydophila psittaci-negative ocular adnexal extranodal marginal zone lymphomas (OAEMZLs) is poorly understood. OAEMZLs are monoclonal tumors expressing a biased repertoire of mutated surface immunoglobulins. Antigenic activation of the B-cell receptor (BCR) may have a role in the
Philipp Müller et al.
Science translational medicine, 7(315), 315ra188-315ra188 (2015-11-27)
Targeted drug delivery with antibody-drug conjugates such as the HER2-directed ado-trastuzumab emtansine (T-DM1) has emerged as a powerful strategy for cancer therapy. We show that T-DM1 is particularly effective in eliciting antitumor immunity in patients with early breast cancer (WSG-ADAPT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service