19-0505
Magnesium sulfate
SAJ first grade (anhydrous), ≥98.0%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade (anhydrous)
form:
solid
Recommended Products
grade
SAJ first grade (anhydrous)
vapor density
<0.01 (vs air)
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
≥98.0%
form
solid
availability
available only in Japan
pH
~7.9 (25 °C, 50 g/L)
SMILES string
[Mg++].[O-]S([O-])(=O)=O
InChI
1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
CSNNHWWHGAXBCP-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological
Robert J Floor et al.
Chembiochem : a European journal of chemical biology, 15(11), 1660-1672 (2014-07-01)
We explored the use of a computational design framework for the stabilization of the haloalkane dehalogenase LinB. Energy calculations, disulfide bond design, molecular dynamics simulations, and rational inspection of mutant structures predicted many stabilizing mutations. Screening of these in small
Lex W Doyle et al.
Obstetrics and gynecology, 113(6), 1327-1333 (2009-05-23)
To systematically review rates of neurologic outcomes reported in childhood for the preterm fetus exposed to antenatal magnesium sulfate. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register, CENTRAL (The Cochrane Library 2008, Issue 3), relevant references from retrieved
A P Morrison et al.
British journal of anaesthesia, 110(5), 702-712 (2013-03-28)
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized
Federica Belluti et al.
ChemMedChem, 9(2), 371-382 (2014-01-10)
A significant improvement in the treatment of trypanosomiases has been achieved with the recent development of nifurtimox-eflornithine combination therapy (NECT). As an alternative to drug combinations and as a means to overcome most of the antitrypanosomatid drug discovery challenges, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service