Skip to Content
Merck
All Photos(1)

Documents

18-0500

Sigma-Aldrich

Lead(II) acetate trihydrate

JIS special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.5%

form

solid

reaction suitability

reagent type: catalyst
core: lead

availability

available only in Japan

mp

75 °C (dec.) (lit.)

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Sobin et al.
Toxicology letters, 220(1), 44-52 (2013-04-20)
The mechanisms by which early chronic lead (Pb) exposure alter brain development have not been identified. We examined neuroimmune system effects in C57BL/6J mice with Pb exposure, including levels that may be common among children in lower socioeconomic income environments.
Mohammed H Badawoud et al.
Neurosciences (Riyadh, Saudi Arabia), 18(1), 52-57 (2013-01-08)
To investigate the morphological changes in the peripheral motor system of the rat induced by exposure to lead. This study was conducted at the Anatomy Department, Faculty of Medicine, King Abdulaziz University, Jeddah, Kingdom of Saudi Arabia from January 2011
Qinghua Jia et al.
Toxicology mechanisms and methods, 22(9), 705-710 (2012-08-17)
Lead-induced nephrotoxicity is a human health hazard problem. In this study, Human mesangial cells (HMCs) were treated with different concentration of lead acetate (5, 10, 20 μmol/l) in order to investigate the oxidative stress and apoptotic changes. It was revealed
Asit Rai et al.
Toxicology mechanisms and methods, 23(2), 99-107 (2012-08-21)
Exposure to a mixture of As, Pb and Cd induces apoptosis and morphological alterations in the cortical astrocytes of rat brain. The levels of the glial fibrillary acidic protein (GFAP) undergo a reduction. The GFAP exists in several isoforms, viz.
Chunping Li et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 225-229 (2013-03-27)
Occupational and environmental exposures to lead (Pb) are a worldwide concern. DNA methylation plays an important role in the development of Pb toxicity. Here, we try to find out the evidence to prove that the methylation of the LINE-1 promoter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service