Skip to Content
Merck
All Photos(1)

Key Documents

15-1800

Sigma-Aldrich

2-Methyl-1-propanol

SAJ first grade, ≥99.0%

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
SAJ first grade
bp:
108 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

2.55 (vs air)

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.396 (lit.)

bp

108 °C (lit.)

mp

−108 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC(C)CO

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Li et al.
Science (New York, N.Y.), 335(6076), 1596-1596 (2012-03-31)
One of the major challenges in using electrical energy is the efficiency in its storage. Current methods, such as chemical batteries, hydraulic pumping, and water splitting, suffer from low energy density or incompatibility with current transportation infrastructure. Here, we report
D Norbäck et al.
Occupational and environmental medicine, 52(6), 388-395 (1995-06-01)
As a part of the worldwide European Community respiratory health survey, possible relations between symptoms of asthma, building characteristics, and indoor concentration of volatile organic compounds (VOCs) in dwellings were studied. The study comprised 88 subjects, aged 20-45 years, from
Arul M Varman et al.
Applied and environmental microbiology, 79(3), 908-914 (2012-11-28)
Global warming and decreasing fossil fuel reserves have prompted great interest in the synthesis of advanced biofuels from renewable resources. In an effort to address these concerns, we performed metabolic engineering of the cyanobacterium Synechocystis sp. strain PCC 6803 to
Jan-Karl Guterl et al.
ChemSusChem, 5(11), 2165-2172 (2012-10-23)
The limited supply of fossil resources demands the development of renewable alternatives to petroleum-based products. Here, biobased higher alcohols such as isobutanol are versatile platform molecules for the synthesis of chemical commodities and fuels. Currently, their fermentation-based production is limited
Pedram Fatehi
Biotechnology progress, 29(2), 297-310 (2013-01-09)
In this review, the recent advancements and technical challenges associated with the production of ethanol, butanol, and isobutanol via bioconversion routes from celluloses of woody materials are reviewed. Physicochemical processes, e.g. steam explosion, seem to be the most viable process

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service