Skip to Content
Merck
All Photos(1)

Key Documents

15-1070

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

JIS special grade, ≥99.0%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

3-5 (20 °C, 50 g/L)

mp

100 °C (dec.) (lit.)

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jung Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 520-524 (2005-04-07)
This study examined whether iron accumulated in ferritin-producing recombinant microbes is bioavailable to rats with iron deficiency. Rats induced with iron deficiency were treated with iron preparations of ferrous ammonium sulfate, horse spleen ferritin, control yeast, and ferritin-producing recombinant yeast
Kazumasa Kumamoto et al.
Free radical research, 39(9), 987-993 (2005-08-10)
Identification of free radicals was performed for the reaction mixtures of autoxidized 1,2-dilinoleoylphosphatidylcholine (DLPC) with ferrous ions (or DLPC hydroperoxide with ferrous ions) and of DLPC with soybean lipoxygenase using electron spin resonance (ESR), high performance liquid chromatography (HPLC)--ESR and
Ubon Cha'on et al.
Toxicological sciences : an official journal of the Society of Toxicology, 96(1), 194-201 (2006-12-19)
The aim of this study is to identify the biochemical mechanism of phosphine toxicity and resistance, using Caenorhabditis elegans as a model organism. To date, the precise mode of phosphine action is unclear. In this report, we demonstrate the following
Y J Tseng et al.
Medical physics, 32(4), 1017-1023 (2005-05-18)
A least-squares error minimization approach was adopted to assess ferric ion diffusion coefficient of Fricke-agarose gels. Ferric ion diffusion process was modeled as a Gaussian-shaped degradation kernel operating on an initial concentration distribution. Diffusion coefficient was iteratively determined by minimizing
Melanie T M Davidson et al.
Medical physics, 36(4), 1340-1350 (2009-05-29)
Granulated cane sugar has been used as a dosimetric material to report dose in high dose accidental irradiations. The purpose of this study was to assess whether clinical dosimetry is also plausible with such a commonly available material. The behavior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service