Skip to Content
Merck
All Photos(1)

Key Documents

13-1940

Sigma-Aldrich

Hydroquinone

SAJ first grade, ≥99.0%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥99.0%
bp:
285 °C (lit.)
vapor pressure:
1 mmHg ( 132 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

Assay

≥99.0%

form

crystalline

autoignition temp.

930 °F

availability

available only in Japan

pH

3.7 (70 g/L)

bp

285 °C (lit.)

mp

172-175 °C (lit.)

solubility

water: soluble 50 g/L

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqi Hu et al.
Biosensors & bioelectronics, 47, 45-49 (2013-04-03)
A novel light "on-off" photoelectrochemical sensing for the sensitive determination of hydroquinone was developed based on the porphyrin-functionalized Au nanoparticles on graphene (porphyrin/AuNPs/graphene). Gold nanoparticles (AuNPs) were firstly modified onto graphene sheets using NaBH4 as reductant and the porphyrin/AuNPs/graphene nanocomposites
P Carbonnelle et al.
Toxicology and applied pharmacology, 132(2), 220-226 (1995-06-01)
Decreased interleukin-1 (IL-1) production by mononuclear phagocytes has been shown to contribute to benzene myelotoxicity in animals. The study presented here was designed to examine the relevance of this mechanism in humans. Fresh human blood monocytes were exposed to 0.001-10
Jacob Levitt
Journal of the American Academy of Dermatology, 57(5), 854-872 (2007-05-01)
Recently, the US Food and Drug Administration proposed a ban on over-the-counter hydroquinone mainly on the basis of high absorption, reports of exogenous ochronosis in humans, and murine hepatic adenomas, renal adenomas, and leukemia with large doses over extended time
Douglas McGregor
Critical reviews in toxicology, 37(10), 887-914 (2007-11-21)
The toxicology of hydroquinone has been reviewed on a number of previous occasions. This review targets its potential for carcinogenicity and possible modes of carcinogenic action. The evaluation made by IARC (1999) of its carcinogenic risk to humans was that
A P DeCaprio
Critical reviews in toxicology, 29(3), 283-330 (1999-06-24)
Hydroquinone (HQ) is a high-volume commodity chemical used as a reducing agent, antioxidant, polymerization inhibitor, and chemical intermediate. It is also used in over-the-counter (OTC) drugs as an ingredient in skin lighteners and is a natural ingredient in many plant-derived

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service