Skip to Content
Merck
All Photos(1)

Key Documents

13-1890

Sigma-Aldrich

Hydrofluoric acid

55%, CP

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
concentration:
55%
grade:
CP

grade

CP

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

form

liquid

availability

available only in Japan

concentration

55%

density

1.15 g/mL at 25 °C (lit.)

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenlin Yvonne Lin et al.
Waste management (New York, N.Y.), 41, 75-84 (2015-04-22)
Accelerated carbonation has been studied as a treatment method for MSW IBA, and the main advantage is that it can shorten the treatment duration from months to days, compared to natural weathering. This study investigated the effect of accelerated carbonation
Yvonne C J Wientjes et al.
Genetics, selection, evolution : GSE, 47, 42-42 (2015-05-09)
Although simulation studies show that combining multiple breeds in one reference population increases accuracy of genomic prediction, this is not always confirmed in empirical studies. This discrepancy might be due to the assumptions on quantitative trait loci (QTL) properties applied
Ian Ford et al.
International journal of cardiology, 184, 163-169 (2015-02-24)
We identified easily obtained baseline characteristics associated with outcomes in patients with chronic heart failure (HF) and elevated heart rate (HR) receiving contemporary guideline-recommended therapy in the SHIFT trial, and used them to develop a prognostic model. We selected the
Faiez Zannad et al.
European journal of heart failure, 17(7), 735-742 (2015-04-29)
Thrombin is a critical element of crosstalk between pathways contributing to worsening of established heart failure (HF). The aim of this study is to explore the efficacy and safety of rivaroxaban 2.5 mg bid compared with placebo (with standard care) after
Philip J Millar et al.
Circulation, 131(5), 459-468 (2014-12-04)
Muscle sympathetic activation in heart failure with reduced ejection fraction (HFrEF) has been attributed, on the basis of multiunit recordings, to attenuated inhibitory feedback from stretch-sensitive cardiopulmonary mechanoreceptors. However, such preparations integrate 2 populations of single units exhibiting directionally opposite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service