Skip to Content
Merck
All Photos(1)

Key Documents

13-1550

Sigma-Aldrich

Hydrazinium sulfate

SAJ first grade, ≥97.0%

Synonym(s):

Hydrazine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2SO4
CAS Number:
Molecular Weight:
130.12
Beilstein:
8778859
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

≥97.0%

form

crystalline

availability

available only in Japan

mp

254 °C (lit.)

SMILES string

NN.OS(O)(=O)=O

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)

InChI key

ZGCHATBSUIJLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Qu et al.
Biosensors & bioelectronics, 42, 430-433 (2012-12-19)
Graphene-nickel nanoparticle hybrid was prepared by the one-step far infrared-assisted reduction of graphene oxide and nickel (II) ions using hydrazine. It was loaded on the surface of a magnetic electrode for electrochemical sensing. The feasibility and performance of the novel
Paul D Rainville et al.
Bioanalysis, 4(23), 2833-2842 (2012-12-12)
Accurate mass based LC-MS combined with statistical analysis is established as a core analytical technology for metabonomic studies. This is primarily due to the specificity, sensitivity and structural elucidation capabilities of the technology. The vast majority of these studies are
Hatem A Abdel-Aziz et al.
Molecules (Basel, Switzerland), 18(2), 2084-2095 (2013-02-08)
Although salicylaldehyde azine (3) was reported in 1985 as the single product of the reaction of ethyl 2-oxo-2H-chromene-3-carboxylate (1) with hydrazine hydrate, we identified another main reaction product, besides 3, which was identified as malono-hydrazide (4). In the last
Leslie D Field et al.
Inorganic chemistry, 52(3), 1570-1583 (2013-01-24)
Treatment of [RuCl(PP(3)(iPr))](+)Cl(-) (PP(3)(iPr) = P(CH(2)CH(2)P(i)Pr(2))(3)) with hydrazine, phenylhydrazine, and methylhydrazine afforded side-on bound hydrazine complexes [RuCl(η(2)-H(2)N-NH(2))(η(3)-PP(3)(iPr))](+), [RuCl(η(2)-H(2)N-NHPh)(η(3)-PP(3)(iPr))](+), and [RuCl(η(2)-H(2)N-NHMe)(η(3)-PP(3)(iPr))](+). The analogous reactions of [RuCl(2)(PP(3)(Ph))] (PP(3)(Ph) = P(CH(2)CH(2)PPh(2))(3)) with hydrazine, phenylhydrazine, and methylhydrazine afforded end-on bound hydrazine complexes [RuCl(η(1)-H(2)N-NH(2))(PP(3)(Ph))](+), [RuCl(η(1)-H(2)N-NHPh)(PP(3)(Ph))](+)
John M Keith
The Journal of organic chemistry, 77(24), 11313-11318 (2012-12-12)
Various pyridine, quinoline, isoquinoline, and pyrimidine N-oxides were converted to their corresponding α-N-aryltriflamidoheteroarenes in good yield by treatment with N-aryltriflimides, both neat and in solution, at temperatures ranging from rt to 100 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service