Skip to Content
Merck
All Photos(1)

Key Documents

12-0820

Sigma-Aldrich

Glucose stock solution

1000 mg/dL

Synonym(s):

D-(+)-Glucose solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1281608
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

availability

available only in Japan

concentration

1000 mg/dL

dilution

(for analytical testing)

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christel Thauvin-Robinet et al.
American journal of human genetics, 93(1), 141-149 (2013-07-03)
Short stature, hyperextensibility of joints and/or inguinal hernia, ocular depression, Rieger anomaly, and teething delay (SHORT) syndrome is a developmental disorder with an unknown genetic cause and hallmarks that include insulin resistance and lack of subcutaneous fat. We ascertained two
Mark-Steven Steiner et al.
Chemical Society reviews, 40(9), 4805-4839 (2011-06-16)
This critical review covers the present state of the art in optical sensing of glucose. Following an introduction into the significance of (continuous) sensing of glucose and a brief look back, we discuss methods based on (a) monitoring the optical
Jocelyn Campbell et al.
Ecology, 94(5), 1186-1195 (2013-07-19)
Foliose lichens with cyanobacterial bionts (bipartite and tripartite) form a distinct assemblage of epiphytes strongly associated with humid microclimatic conditions in inland British Columbia. Previous research showed that these cyano- and cephalolichen communities are disproportionately abundant and species-rich on conifer
David Cruz-Garcia et al.
The Journal of cell biology, 207(6), 695-703 (2014-12-17)
Upon starvation, Grh1, a peripheral membrane protein located at endoplasmic reticulum (ER) exit sites and early Golgi in Saccharomyces cerevisiae under growth conditions, relocates to a compartment called compartment for unconventional protein secretion (CUPS). Here we report that CUPS lack
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service