Skip to Content
Merck
All Photos(1)

Key Documents

11-0720

Sigma-Aldrich

Formaldehyde solution

JIS special grade, 36.0-38.0%, contains methanol as stabilizer

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
41115710
PubChem Substance ID:
grade:
JIS special grade
application(s):
sample preparation
bp:
97 °C ((37% solution) (lit.))
vapor pressure:
52 mmHg ( 37 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

methanol as stabilizer

availability

available only in Japan

concentration

36.0-38.0%

dilution

(for analytical testing)

bp

97 °C ((37% solution) (lit.))

mp

-15 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

application(s)

sample preparation

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES
Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
Silvia Surinova et al.
EMBO molecular medicine, 7(9), 1166-1178 (2015-08-09)
Non-invasive detection of colorectal cancer with blood-based markers is a critical clinical need. Here we describe a phased mass spectrometry-based approach for the discovery, screening, and validation of circulating protein biomarkers with diagnostic value. Initially, we profiled human primary tumor
James Larkin et al.
The New England journal of medicine, 373(1), 23-34 (2015-06-02)
Nivolumab (a programmed death 1 [PD-1] checkpoint inhibitor) and ipilimumab (a cytotoxic T-lymphocyte-associated antigen 4 [CTLA-4] checkpoint inhibitor) have been shown to have complementary activity in metastatic melanoma. In this randomized, double-blind, phase 3 study, nivolumab alone or nivolumab plus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service