Skip to Content
Merck
All Photos(1)

Key Documents

09-1940

Sigma-Aldrich

2-Ethyl-1-hexanol

SAJ first grade, ≥99.0%

Synonym(s):

Isooctyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719280
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.49 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

550 °F

expl. lim.

0.88 %, 104 °F
9.7 %, 113 °F

availability

available only in Japan

refractive index

n20/D 1.431 (lit.)

bp

183-186 °C (lit.)

mp

−76 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)CO

InChI

1S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3

InChI key

YIWUKEYIRIRTPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandro Nalli et al.
Environmental pollution (Barking, Essex : 1987), 140(1), 181-185 (2005-08-30)
2-Ethylhexanol has been identified as a volatile organic compound (VOC) that contributes to the deterioration of indoor air quality. Plasticizers are common components of dust and building materials and are shown to be degraded by a variety of bacteria and
Gunilla Wieslander et al.
Archives of environmental & occupational health, 65(1), 3-11 (2010-02-12)
The authors evaluated changes of symptoms and biomarkers in health care staff (N = 18) for people with different physical dysfunctions and similarly in an external office control group in a nondamp building (N = 15). The first workplace had
L Ernstgård et al.
Indoor air, 20(2), 168-175 (2010-04-23)
The objective was to assess acute effects from controlled exposure of volunteers to 2-ethyl-1-hexanol, a volatile organic compound that is often found in indoor air. Sixteen males and fourteen females were in random order exposed to 1 mg/m(3) of vapors
D Fragiadakis et al.
The Journal of chemical physics, 132(14), 144505-144505 (2010-04-22)
The dielectric spectra of most simple liquids are characterized by two relaxation processes: (i) the alpha-process, an intense, broad non-Debye relaxation with a non-Arrhenius temperature dependence and (ii) a beta process, evident mainly below the glass transition and having nearly
Christoph van Thriel et al.
Neurotoxicology, 28(2), 347-355 (2006-05-23)
Historically, acute solvent neurotoxicity was strongly related to reversible narcotic states that could be detected by neurobehavioral tests (e.g., simple reaction time). Nowadays, the occupational exposure to chemicals is markedly reduced and the avoidance of chemosensory effects is more important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service