Skip to Content
Merck
All Photos(1)

Documents

09-1500

Sigma-Aldrich

Ethylenediaminetetraacetic acid iron(III) sodium salt

SAJ special grade, ≥95.0%

Synonym(s):

EDTA iron(III) sodium salt, Edathamil, Komplexon II

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2NaFeO8
CAS Number:
Molecular Weight:
367.05
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ special grade

Assay

≥95.0%

form

crystalline

availability

available only in Japan

mp

80 °C

solubility

water: 72 g/L at 25 °C

shipped in

ambient

SMILES string

[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Fe.Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;+3;+1/p-4

InChI key

MKWYFZFMAMBPQK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. Iron-EDTA enhances damage due to reactive oxygen species, such as depletion of reduced glutathione, formation of methemoglobin, and lipid peroxidation.

Legal Information

Komplexon is a trademark of CU Chemie Uetikon AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Troesch et al.
The Journal of nutrition, 141(5), 822-827 (2011-03-25)
The absorption profile of iron fortificants may be a determinant of their ability to generate nontransferrin-bound iron (NTBI) and, thus, their potential safety. Ferrous iron may be absorbed more rapidly than chelated ferric iron, but differences at the fortification level
Catherine W Macharia-Mutie et al.
The Journal of nutrition, 142(9), 1756-1763 (2012-07-20)
Few studies have evaluated the impact of fortification with iron-rich foods such as amaranth grain and multi-micronutrient powder (MNP) containing low doses of highly bioavailable iron to control iron deficiency anemia (IDA) in children. We assessed the efficacy of maize
Tarun Gera et al.
The American journal of clinical nutrition, 96(2), 309-324 (2012-07-05)
The utility of iron fortification of food to improve iron deficiency, anemia, and biological outcomes is not proven unequivocally. The objectives were to evaluate 1) the effect of iron fortification on hemoglobin and serum ferritin and the prevalence of iron
Zhenyu Yang et al.
Maternal & child nutrition, 7 Suppl 3, 123-128 (2011-10-05)
As a highly bioavailable iron compound, sodium iron (iii) ethylenediaminetetraacetate (NaFeEDTA) has been recommended as a food additive for fortification. The amount of a food additive that can be ingested daily over a lifetime without appreciable health risk is termed
Monica N Orozco et al.
Annals of nutrition & metabolism, 60(2), 108-114 (2012-03-15)
In any context of iron supplementation in the prenatal prophylaxis or therapeutic dosage range, a large amount will remain unabsorbed and pass through the intestinal tract into the colonic digesta possibly causing increased oxidation. To compare the generation of fecal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service