Skip to Content
Merck
All Photos(1)

Documents

07-2770

Sigma-Aldrich

Diethylamine

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.5 (vs air)

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

594 °F

expl. lim.

10.1 %

availability

available only in Japan

refractive index

n20/D 1.385 (lit.)

pH

13 (20 °C, 100 g/L)

bp

55 °C (lit.)

mp

−50 °C (lit.)

density

0.707 g/mL at 25 °C (lit.)

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doron Yariv et al.
Colloids and surfaces. B, Biointerfaces, 78(2), 185-192 (2010-04-07)
In this paper we examined feasible correlations between the structure of different lyotropic mesophases and transdermal administration of three diclofenac derivatives with varying degrees of kosmotropic or chaotropic properties, solubilized within the mesophases. It was found that the most chaotropic
Jun Guo et al.
Journal of chromatography. A, 1307, 66-72 (2013-08-15)
The extraction efficiency of analytes in a cold fiber headspace solid-phase microextraction (CF-HS-SPME) system was investigated both theoretically and experimentally. The system was applied for quantitative extraction of polycyclic aromatic hydrocarbons (PAHs) from solid matrices. In order to achieve better
Roberto Cirilli et al.
Analytical chemistry, 81(9), 3560-3570 (2009-03-28)
Second-order rate constants of the diethylamine-promoted enantiomerization of 2-[2-(1-methyl-1H-pyrrol-2-yl)-2-oxo-1-phenylethyl]-isoindole-1,3-dione, a chiral alpha-substituted ketone endowed with high anti-MAO activity type-A, were measured by dynamic high-performance liquid chromatography (DHPLC), stopped-flow high-performance liquid chromatography (sf-HPLC), and a classical method based on enantioselective HPLC
Rakesh Naduvile Veedu et al.
The journal of physical chemistry. A, 112(40), 9742-9750 (2008-09-16)
Five aryliminopropadienones 4a- d have been synthesized by flash vacuum thermolysis (FVT) by using two different precursors in each case. These compounds were deposited at 50 K at a pressure of ca. 10(-6) mbar together with three different nucleophiles, namely
L Rubio et al.
International journal of pharmaceutics, 386(1-2), 108-113 (2009-11-20)
This work evaluates the effect of different bicellar systems on the percutaneous absorption of diclofenac diethylamine (DDEA) using two different approaches. In the first case, the drug was included in bicellar systems, which were applied on the skin and, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service