Skip to Content
Merck
All Photos(1)

Documents

07-1470

Sigma-Aldrich

2,6-Dibromoquinone-4-chloroimide

JIS special grade

Synonym(s):

2,6-Dibromo-N-chloro-p-benzoquinoneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Br2ClNO
CAS Number:
Molecular Weight:
299.35
Beilstein:
2094249
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

JIS special grade

form

solid

availability

available only in Japan

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cl\N=C1\C=C(Br)C(=O)C(Br)=C1

InChI

1S/C6H2Br2ClNO/c7-4-1-3(10-9)2-5(8)6(4)11/h1-2H

InChI key

JYWKEVKEKOTYEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Self-react. A - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Post-column derivatization of vitamin B6 using 2,6-dibromoquinone-4-chlorimide.
T Kawamoto et al.
Journal of chromatography, 267(2), 414-419 (1983-09-16)
C L Garganta et al.
Analytical biochemistry, 240(2), 177-184 (1996-09-05)
Lipoic acid is a coenzyme for pyruvate dehydrogenase, alpha-ketoglutarate dehydrogenase, branched chain-ketoacid dehydrogenase, and the glycine cleavage system. Lipoic acid is covalently attached through an amide to the epsilon-amino group of specific lysine residues of these enzymes. Lipoamidase hydrolyzes the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service