Skip to Content
Merck
All Photos(1)

Documents

05-0300

Sigma-Aldrich

Cadmium sulfate 8/3-hydrate

SAJ super special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4 · 8/3H2O
CAS Number:
Molecular Weight:
256.57
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ super special grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

availability

available only in Japan

SMILES string

[Cd++].[Cd++].[Cd++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3Cd.3H2O4S.8H2O/c;;;3*1-5(2,3)4;;;;;;;;/h;;;3*(H2,1,2,3,4);8*1H2/q3*+2;;;;;;;;;;;/p-6

InChI key

AGHQMAQBLOTWPQ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shurong Tang et al.
Biosensors & bioelectronics, 42, 608-611 (2012-12-25)
A novel highly sensitive and selective electrochemical sensing system based on DNAzyme and rolling circle amplification (RCA) for the determination of Pb²⁺ was developed in the present study. Firstly, the DNAzyme catalytic strands were immobilized onto magnetic beads surface and
Yun-Jie Yang et al.
Acta biomaterialia, 5(9), 3488-3496 (2009-05-16)
Mesoporous silica nanotubes (MSNTs) and amine-functionalized MSNTs (NH(2)-MSNTs) have been successfully synthesized via a sol-gel route using needle-like CaCO(3) nanoparticles as inorganic templates and post-modification with 3-aminopropyltriethoxysilane. Subsequently, the preformed nanotubes were functionalized with blue fluorescent CdS quantum dots, as
Renguo Xie et al.
Journal of the American Chemical Society, 131(42), 15457-15466 (2009-09-25)
The initial formation of semiconductor nanocrystals/nanoclusters, that is, nucleation in the classic literature, was examined both theoretically and experimentally. An experimental method based on determining the initial reaction rate for the formation of nanocrystals/nanoclusters with fixed size and size distribution
Els Keunen et al.
Plant physiology and biochemistry : PPB, 49(10), 1084-1091 (2011-08-20)
Plants exposed to cadmium (Cd) show morphological and physiological disorders. To increase our knowledge regarding Cd-induced signalling, most often the effects of acute exposure are investigated. However, this does not allow in-depth analysis of morphological effects. Therefore, we chronically exposed
Electric-field enhancement of photovoltaic devices: a third reason for the increase in the efficiency of photovoltaic devices by carbon nanotubes.
Wonjoo Lee et al.
Advanced materials (Deerfield Beach, Fla.), 22(20), 2264-2267 (2010-03-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service