Skip to Content
Merck
All Photos(1)

Documents

05-0280

Sigma-Aldrich

Cadmium standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Cd+2, 1000 ppm Cd+2

Synonym(s):

Cadmium nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
8137359
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Cd+2
1000 ppm Cd+2

suitability

suitable for atomic absorption spectrometry

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Target Organs

Kidney,Bone

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

O A Rozentsvet et al.
Izvestiia Akademii nauk. Seriia biologicheskaia, (2)(2), 232-239 (2005-07-12)
The effect of cadmium on clapsinng-leaved pondweed (Potamogeton perfoliatus L.) within the concentration range of 1-1000 microM was studied. It was shown that P. perfoliatus leaves accumulated cadmium during three days. This process was accompanied by changes in leaf morphology.
M S Monteiro et al.
Ecotoxicology and environmental safety, 72(3), 811-818 (2008-10-28)
Laboratory and field studies have provided encouraging insights into the capacity of plants to act as biomonitors of environmental quality through the use of biomarkers. However, a better understanding of the overall process of Cd-induced senescence, describing the cascade of
Effects of cadmium and lead on phytochelatin accumulation in maize shoots and different root parts.
I V Seregin et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 415, 304-306 (2007-10-13)
Emi Dote et al.
Journal of occupational health, 49(1), 17-24 (2007-02-23)
Cadmium nitrate Cd(NO(3))(2) (CdN) is commonly used in Ni-Cd battery factories. The possibility of accidental exposure to CdN is great. CdN is very soluble in water compared to other Cd compounds. Therefore, acute toxicity would be expected to be quick
G De Boeck et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 155(4), 503-508 (2010-01-02)
Recent studies indicate that elasmobranch fish respond differently to metal exposure than marine teleosts. Accumulation rates can be high, which despite the fact that normal background levels for metals in the marine environment are low, is worrying due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service