Skip to Content
Merck
All Photos(1)

Key Documents

03-2350

Sigma-Aldrich

Biotin

SAJ special grade, ≥98.0%

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
41116133
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

powder

availability

available only in Japan

color

white

mp

231-233 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart Trenholm et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(37), 14927-14938 (2013-09-13)
Recently, we demonstrated that gap junction coupling in the population of superior coding ON-OFF directionally selective ganglion cells (DSGCs) genetically labeled in the Hb9::eGFP mouse retina allows the passage of lateral anticipatory signals that help track moving stimuli. Here, we
Konstantinos Ampatzis et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10875-10886 (2013-06-28)
In vertebrates, spinal circuits drive rhythmic firing in motoneurons in the appropriate sequence to produce locomotor movements. These circuits become active early during development and mature gradually to acquire the flexibility necessary to accommodate the increased behavioral repertoire of adult
A Limat et al.
Archives of dermatological research, 288(1), 31-38 (1996-01-01)
In humans and in animals, biotin deficiency causes pathological changes in the skin and its appendages. High doses of biotin may also have beneficial effects on skin, hair and fingernails in humans and animals with normal biotin status. Therefore, we
Adeline Huiling Loo et al.
Nanoscale, 5(17), 7844-7848 (2013-07-13)
Graphene related materials have been widely employed as highly efficient transducers for biorecognition. Here we show a conceptually new approach of using graphene oxide nanoplatelets (50 × 50 nm) as voltammetric inherently active labels for specific protein-target molecule recognition. This
Tuan V Bui et al.
Neuron, 78(1), 191-204 (2013-04-16)
Accurate motor performance depends on the integration in spinal microcircuits of sensory feedback information. Hand grasp is a skilled motor behavior known to require cutaneous sensory feedback, but spinal microcircuits that process and relay this feedback to the motor system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service