Skip to Content
Merck
All Photos(1)

Key Documents

01-4560

Sigma-Aldrich

Ammonium chloride

SAJ first grade, ≥98.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

Assay

≥98.5%

form

crystalline powder

availability

available only in Japan

pH

4.5-5.5 (20 °C, 50 g/L)

mp

340 °C (subl.) (lit.)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Burkard et al.
PloS one, 9(7), e101762-e101762 (2014-07-16)
Studies of viral entry into host cells often rely on the detection of post-entry parameters, such as viral replication or the expression of a reporter gene, rather than on measuring entry per se. The lack of assays to easily detect
Allison L Boyd et al.
The Journal of experimental medicine, 211(10), 1925-1935 (2014-09-03)
Allogeneic hematopoietic stem cell (HSC) transplantation (HSCT) is currently the leading strategy to manage acute myeloid leukemia (AML). However, treatment-related morbidity limits the patient generalizability of HSCT use, and the survival of leukemic stem cells (LSCs) within protective areas of
Cyrill A Rentsch et al.
European urology, 66(4), 677-688 (2014-03-29)
Whether the commonly used bacillus Calmette-Guérin (BCG) strains Connaught and Tice confer different treatment responses in non-muscle-invasive bladder cancer (NMIBC) is unknown. To compare clinical efficacy, immunogenicity, and genetics of BCG Connaught and Tice. A prospective randomized single-institution trial with
Sewa Rijal et al.
Blood, 125(18), 2815-2824 (2015-03-05)
Phosphoinositide signaling regulates diverse cellular functions. Phosphoinositide-3 kinase (PI3K) generates PtdIns(3,4,5)P3 and PtdIns(3,4)P2, leading to the activation of proliferative and anti-apoptotic signaling pathways. Termination of phosphoinositide signaling requires hydrolysis of inositol ring phosphate groups through the actions of PtdIns(3,4,5)P3 3-phosphatase
Chloé Lescale et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(2), 455-463 (2014-11-08)
Within the bone marrow, the endosteal niche plays a crucial role in B-cell differentiation. Because spaceflight is associated with osteoporosis, we investigated whether changes in bone microstructure induced by a ground-based model of spaceflight, hind limb unloading (HU), could affect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service