Skip to Content
Merck
All Photos(1)

Documents

01-3110

Sigma-Aldrich

Aluminon

JIS special grade

Synonym(s):

Aurintricarboxylic acid ammonium salt, ATA, Aluminon, Ammonium aurintricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H14O9 · 3NH3
CAS Number:
Molecular Weight:
473.43
Colour Index Number:
43810
Beilstein:
3900820
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

JIS special grade

form

powder

availability

available only in Japan

mp

220-225 °C (dec.) (lit.)

solubility

water: soluble

shipped in

ambient

storage temp.

room temp

SMILES string

N.N.N.OC(=O)c1cc(ccc1O)\C(c2ccc(O)c(c2)C(O)=O)=C3/C=CC(=O)C(=C3)C(O)=O

InChI

1S/C22H14O9.3H3N/c23-16-4-1-10(7-13(16)20(26)27)19(11-2-5-17(24)14(8-11)21(28)29)12-3-6-18(25)15(9-12)22(30)31;;;/h1-9,23-24H,(H,26,27)(H,28,29)(H,30,31);3*1H3

InChI key

AIPNSHNRCQOTRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Aurintricarboxylic acid readily polymerizes in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

Other Notes

May contain a substantial amount of polymeric material.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Chen Hung et al.
The Journal of antimicrobial chemotherapy, 65(4), 676-683 (2010-01-22)
Enterovirus 71 (EV71) causes serious diseases in humans. The aim of this study was to examine the effects of aurintricarboxylic acid (ATA) on EV71 replication and to explore the underlying mechanism. To measure the activity of ATA in inhibiting the
Hui-Chen Hung et al.
Antiviral research, 81(2), 123-131 (2008-11-19)
There is a continuing threat that the highly pathogenic avian influenza virus will cause future influenza pandemics. In this study, we screened a library of compounds that are biologically active and structurally diverse for inhibitory activity against influenza neuraminidase (NA).
Jakyung Yoo et al.
Advances in protein chemistry and structural biology, 87, 219-247 (2012-05-23)
DNA methyltransferases (DNMTs) are promising epigenetic targets for the development of novel anticancer drugs and other diseases. Molecular modeling and experimental approaches are being used to identify and develop inhibitors of human DNMTs. Most of the computational efforts conducted so
Qiang Peng et al.
PloS one, 7(10), e47273-e47273 (2012-11-03)
Baboons receiving xenogeneic livers from wild type and transgenic pigs survive less than 10 days. One of the major issues is the early development of profound thrombocytopenia that results in fatal hemorrhage. Histological examination of xenotransplanted livers has shown baboon
Moonhee Lee et al.
Neurobiology of aging, 33(10), 2237-2246 (2012-01-06)
Complement plays a vital role in both the innate and adaptive immune systems. It recognizes a target, opsonizes it, generates anaphylatoxins, and directly kills cells through the membrane attack complex (MAC). This final function, which assembles C5b-9(n) on viable cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service