Skip to Content
Merck
All Photos(1)

Documents

01-1700

Sigma-Aldrich

Adipic acid

SAJ special grade, ≥99.5%

Synonym(s):

Hexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1209788
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 159.5 °C)

Assay

≥99.5%

form

solid

autoignition temp.

788 °F

availability

available only in Japan

bp

265 °C/100 mmHg (lit.)

mp

151-154 °C (lit.)

SMILES string

OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roohan Rakhshaee et al.
Journal of hazardous materials, 189(1-2), 158-166 (2011-03-15)
The removal of methylene blue (MB) as a cationic dye from aqueous solution by the stabilized Fe(3)O(4) nano-particles with the extracted pectin from apple waste (FN-PA) increased due to using the cross-linked forms of the bound pectin on the nano-particles
H V Pawar et al.
Colloids and surfaces. B, Biointerfaces, 102, 102-110 (2012-09-26)
Streptomycin (STP) and diclofenac (DLF) loaded film dressings were prepared by blending Polyox(®) (POL) with four hydrophilic polymers [hydroxypropylmethylcellulose (HPMC), carrageenan (CAR), sodium alginate (SA) or chitosan (CS)] using glycerol (GLY) as plasticiser. The films were characterised by scanning electron
Holger Noack et al.
Inorganic chemistry, 50(4), 1194-1202 (2011-01-28)
Adipic acid is a key compound in the chemical industry, where it is mainly used in the production of polymers. The conventional process of its generation requires vast amounts of energy and, moreover, produces environmentally deleterious substances. Thus, there is
Tânia Veiga et al.
Metabolic engineering, 14(4), 437-448 (2012-04-25)
Industrial production of semi-synthetic cephalosporins by Penicillium chrysogenum requires supplementation of the growth media with the side-chain precursor adipic acid. In glucose-limited chemostat cultures of P. chrysogenum, up to 88% of the consumed adipic acid was not recovered in cephalosporin-related
Krzysztof J Paluch et al.
Journal of pharmaceutical sciences, 100(8), 3268-3283 (2011-04-08)
The production of salt or cocrystalline forms is a common approach to alter the physicochemical properties of pharmaceutical compounds. The goal of this work was to evaluate the impact of anion choice (succinate, adipate, and sulfate) on the physicochemical characteristics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service