TAQNTKB
Roche
KAPA Taq PCR Kit with dNTPs
Synonym(s):
PCR kit, taq
About This Item
Recommended Products
shelf life
≤18 mo.
Quality Level
feature
dNTPs included
hotstart: no
packaging
pkg of 250 U (KK1008)
pkg of 2500 U (BK1001)
pkg of 500 U (KK1016)
pkg of 5000 U (BK1003)
manufacturer/tradename
Roche
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
- High throughput PCR
- Amplification of low copy DNA templates
- Multiplex PCR
- Specific amplification of complex templates
- RT-PCR
- Quantitative polymerase chain reaction (qPCR)
Biochem/physiol Actions
Features and Benefits
- Improved sensitivity, specificity, and yields.
- Novel buffer formulation facilitates specific primer annealing, leading to higher yield of specific product.
Quick Notes:
- KAPA Taq HotStart® DNA Polymerase can replace any commercial hotstart Taq DNA polymerase in an existing protocol. The final MgCl2 concentration and annealing temperature may need to be optimized to account for differences in formulation.
- The KAPA Taq HotStart Buffer is a uniquelyformulated buffer offering improved specificity and sensitivity, and improved amplification of GC- and AT-rich templates.
- The KAPA Taq HotStart Buffer does not contain MgCl2; MgCl2 (25 mM) is supplied separately to allow greater flexibility during reaction setup.
- The KAPA Taq HotStart PCR Kit is suitable for the amplification of fragments up to 3.5 kb from genomic DNA or 5 kb from less complex targets.
Quality
Preparation Note
Other Notes
Legal Information
Kit Components Only
- KAPA Taq DNA Polymerase 5 U/µL
- 10X KAPA Taq Buffer A
- 10X KAPA Taq Buffer B
- MgCl2 25 mM
- KAPA dNTP Mix 10 mM each
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service