Skip to Content
Merck
All Photos(3)

Key Documents

LRHSNTKB

Roche

KAPA LongRange

HotStart PCR Kit

Synonym(s):

PCR

Sign Into View Organizational & Contract Pricing

Select a Size

250 U
$318.00
500 U
$591.00

$318.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
250 U
$318.00
500 U
$591.00

About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

$318.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

usage

sufficient for 150 reactions
sufficient for 400 reactions
sufficient for 800 reactions

shelf life

≤18 mo.

feature

High Fidelity PCR
Long & Accurate PCR
dNTPs included
hotstart

packaging

pkg of 100 U (KK3501)
pkg of 250 U (KK3502)
pkg of 500 U (KK3503)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The KAPA Long Range PCR system is a blend of Taq DNA polymerase and an engineered archaeal (B-family) DNA polymerase possessing proofreading capability. This two-enzyme system is designed specifically to support long-range and/or sensitive PCR, and possess 3 – 4X higher fidelity than Taq polymerase. In the hot-start formulation, KAPA Long Range is combined with a proprietary antibody that inactivates the enzyme until the first denaturation step, eliminating spurious amplification products and increasing reaction efficiency and sensitivity.

KAPA Long Range is also supplied in 2X ReadyMix format with loading dye that contains all the components required for PCR except primers and template. The loading dye in the reaction buffer allows you to load PCR products directly onto the agarose gel with no extra steps.

Application

KAPA LongRange has been used in:
  • Amplification of long targets and/or low amounts
  • Standard short- and mid-range PCR
  • T-A cloning
  • Targeted sample enrichment for next-generation DNA sequencing
  • polymerase chain reaction (PCR)[1][2]

Biochem/physiol Actions

Both KAPA Taq HotStart® DNA polymerase and a modified archaeal (Type B) DNA polymerase enzyme possess 5′→3′ polymerase activity, but only KAPA Taq HotStart DNA polymerase possesses 5′→3′ exonuclease activity and only the Type B DNA polymerase possesses 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA LongRange HotStart is 2–4 fold higher than that of Taq DNA Polymerase, but is lower than the fidelity of pure proofreading polymerases such as KAPA HiFi. Reaction products generated with KAPA LongRange HotStart are polyA-tailed and can be cloned into TA-cloning vectors.

Features and Benefits

Amplify long targets with high sensitivity:
  • Amplification of targets with high sensitivity and specificity up to 15 kb
  • High yields with low input DNA

Outperforms with complex targets:
  • Exhibits high yield, greater sensitivity and specificity where competitor long-range systems fail

Quick Notes:
  • KAPA LongRange HotStart is a blend of KAPA Taq HotStart and a proofreading DNA polymerase.
  • Suitable for short-, medium- and long-range PCR, and PCR with low amounts of template DNA.
  • Fidelity is 2–4 times better than Taq.For short amplicons, replace Taq DNA polymerase for improvements in yield and/or fidelity.
  • Amplify mid- to long-range targets with high yield and sensitivity.
  • KAPA LongRange Buffer (Mg-free) is designed for long-range, sensitive PCR. MgCl2 is supplied separately for optimization.

Quality

Each batch of KAPA LongRange HotStart DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). The KAPA LongRange HotStart PCR system is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for longterm storage.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.

Kit Components Only

Product No.
Description

  • KAPA Long Range HotStart® DNA Polymerase 5 U/μL or 2.5 U/μL

  • 5X KAPA Long Range Buffer

  • MgCl2 25 mM

  • dNTP Mix 10 mM each dNTP

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ameet S Sengar et al.
Cell reports, 29(13), 4285-4294 (2019-12-26)
NMDA receptors (NMDARs) are critical for physiological synaptic plasticity, learning, and memory and for pathological plasticity and neuronal death. The GluN1 subunit is encoded by a single gene, GRIN1, with 8 splice variants, but whether the diversity generated by this
Martyna Lukoseviciute et al.
Developmental cell, 47(5), 608-628 (2018-12-05)
The neural crest (NC) is a transient embryonic stem cell-like population characterized by its multipotency and broad developmental potential. Here, we perform NC-specific transcriptional and epigenomic profiling of foxd3-mutant cells in vivo to define the gene regulatory circuits controlling NC specification. Together

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service