Skip to Content
Merck

Skip To

LIBDL-RO

Roche

Liberase DL Research Grade

low Dispase concentration

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200


form

lyophilized

packaging

pkg of 100 mg (05466202001 [2 x 50 mg]), pkg of 10 mg (05401160001 [2 x 5 mg])

manufacturer/tradename

Roche

parameter

35-37 °C optimum reaction temp.

optimum pH

7.4

shipped in

dry ice

storage temp.

−20°C

General description

Liberase DH Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a high concentration of Dispase®, a non-clostridial neutral protease. Liberase DL (Dispase Low) Research Grade is used for the dissociation of a broad range of tissue types, where high purity of the enzyme blend is necessary for high cell yield and viability. Bacterial by-products, such as endotoxins, are reduced up to several thousand-fold.

Application

Liberase DL (Dispase Low) Research Grade has been used for the isolation of skin stem cells from mice.[1] It is also used for the dissociation of a broad range of tissue types, where high purity of the enzyme blend is necessary for high cell yield and viability. Bacterial by-products, such as endotoxins, are reduced up to several thousand-fold.

Features and Benefits

  • Maximize viability and yield of isolated cells with an enzyme blend that has less clostripain and trypsin activity, as well as reduced endotoxin content.
  • Count on higher specific activity of the enzyme blend as a result of higher Collagenase I + II purity (determined by HPLC analysis).
  • Obtain higher experimental reproducibility due to higher lot-to-lot consistency.

Preparation Note

Stabilizers: Calcium
Working concentration: Liberase Research Grade Enzyme Working Concentration

Liberase enzymes have significantly higher specific activities than traditional collagenases. This means that the working concentration of Liberase Research Grade Purified Enzymes, expressed in mg/ml, will be lower than that of traditional collagenase.

When the application is on the Roche list of applications at www.collagenase.com, use the Liberase Research Grade concentration recommended for that application.

When the application is not included on this list, first use Liberase TM Research Grade at a concentration of 0.08–0.28 Wünsch units/ml.

The goal is to determine the best starting concentration of Liberase Research Grade Enzyme Blends. This is a starting point, and the final concentration may vary due to differences in procedure and lot-to-lot differences in traditional collagenase.

Collagenase Working Concentration

Multiply your previous collagenase working concentration (mg/ml) by its specific activity (Wünsch units/mg, [as determined above]), to obtain Wünsch units/ml. To determine how much Liberase Research Grade Enzyme Blend to use, first multiply your collagenase working concentration (in Wünsch units/ml) times the total volume of your working enzyme solution to obtain the total collagenase activity needed (Wünsch units). Divide the total collagenase activity required by the Liberase Research Grade stock concentration ( “Reconstitution and Storage”). This indicates how many milliliters of Liberase Research Grade Enzyme Blend stock solution to use in your working enzyme solution.
Storage conditions (working solution): Store unused stock solution in single-use aliquots at -15 to -25 °C. For further information on product stability, please visit the Roche Liberase Enzyme website at www.collagenase.com.

Note: Avoid repeated freezing and thawing!
Reconstitute the lyophilized enzyme with injection-quality sterile water or tissue-dissociation buffer. Do not add serum or other components that may influence enzyme activity, such as albumin or protease inhibitors, to the dissociation buffer. Enzyme stability is reduced at higher concentrations and warmer temperatures (4 °C). Avoid both the above conditions.

Reconstitute the entire vial. Do not weigh individual aliquots of the lyophilizate. The introduction of moisture into the vial results in a decline in enzymatic activity.

Place vial on ice to rehydrate the lyophilized enzyme.

Gently agitate the vial at 2 to 8 °C until enzyme is completely dissolved (max. 30 min).

Depending on the type of tissue-dissociation buffer used to dissolve Liberase Research Grade Purified Enzyme Blends, slight precipitations may be observed readily dissolve in the diluted working solution and have no influence on enzyme activity.

Remove an aliquot of the stock solution to prepare the working solution.

Reconstitution volume

2 ml (1 vial with 5 mg–10 mg pack size)
10 ml (1 vial with 50 mg–100 mg pack size)

Collagenase Wünsch (units/ml)

13 (1 vial with 5 mg–10 mg pack size)
26 (1 vial with 50 mg–100 mg pack size)

Total Collagenase concentration [mg/ml]

2.5 (1 vial with 5 mg–10 mg pack size)
5.0 (1 vial with 50 mg–100 mg pack size)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Dispase is a registered trademark of Godo Shusei Co., Ltd.
Liberase is a trademark of Roche


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

Your current tissue dissociation protocol, and the extent to which you have been successful with traditional collagenase are good guides for selecting the appropriate Liberase Enzyme Blend.

Articles

Enzyme Explorer Key Resource: Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Related Content

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.


Maria Fernanda Forni et al.
PloS one, 10(10), e0140143-e0140143 (2015-10-16)
The skin is a rich source of readily accessible stem cells. The level of plasticity afforded by these cells is becoming increasingly important as the potential of stem cells in Cell Therapy and Regenerative Medicine continues to be explored. Several