Skip to Content
Merck
All Photos(1)

Key Documents

ENDOARGS-RO

Roche

Endoproteinase Asp-N Sequencing Grade

from Pseudomonas fragi

Synonym(s):

protease

Sign Into View Organizational & Contract Pricing

Select a Size

2 μG
$238.00
3 X 2 μG
$647.00

$238.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
2 μG
$238.00
3 X 2 μG
$647.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

$238.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Assay

≥90% (SDS-PAGE)

form

lyophilized

specific activity

≥20000 units/mg protein

mol wt

27 kDa

packaging

pkg of 2 μg (11420488001)
pkg of 3 × 2 μg (11054589001)

manufacturer/tradename

Roche

optimum pH

7.0-8.0

pH range

6.5-9.0

storage temp.

2-8°C

General description

The wild type of Pseudomonas fragi segregates a metalloprotease, which cleaves peptide bonds N-terminally at small hydrophilic amino acids. A mutant of this strain produces the Endoproteinase Asp-N Sequencing Grade, which is isolated as a highly purified and specific protease.

Specificity

Endoproteinase Asp-N Sequencing Grade is a metalloprotease. In phosphate, acetate, or Tris buffers at pH 6.0–8.5, it specifically cleaves peptide bonds N-terminally at aspartic and cysteic acid. If cysteine is reduced or alkylated, only -↓-Asp-X is cleaved.
The specificity of Endoproteinase Asp-N Sequencing Grade is verified with glucagon as a substrate. High concentrations of Endoproteinase Asp-N Sequencing Grade (1 part by weight enzyme with 10 parts by weight glucagon) are incubated to detect traces of impurities like contaminating proteases.
Heat inactivation: The protease is stopped by boiling for 5 minutes.

Application

Use Endoproteinase Asp-N Sequencing Grade for protein structure analysis and sequence analysis.[1][2]

Preparation Note

Working concentration: 2 μg /50 μl double-dist. water
One vial (= 2 μg enzyme) is dissolved in 50 μl double-dist. water resulting in a buffer concentration of 10 mM Tris-HCl pH 7.5.
Working solution: Recommended solvent is double-distilled water.
Storage conditions (working solution): -15 to -25 °C
A solution in double-dist. water is stable for 1 week at maximum at 2 to 8 °C and for about 1 month at -15 to -25 °C; repeated thawing should be avoided.

Reconstitution

Lyophilized Endoproteinase Asp-N sequencing grade is reconstituted in 50 μl double-dist. water. This results in a buffer concentration of 10 mM Tris-HCl, pH 7.5. In order to avoid autolysis the incubation temperature should not exceed 37 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Fernandez-Martinez et al.
The Journal of cell biology, 196(4), 419-434 (2012-02-15)
The nuclear pore complex (NPC) is a multiprotein assembly that serves as the sole mediator of nucleocytoplasmic exchange in eukaryotic cells. In this paper, we use an integrative approach to determine the structure of an essential component of the yeast
Ming-Ming Li et al.
Nature communications, 4, 1832-1832 (2013-05-16)
Regulation of actomyosin dynamics by post-transcriptional modifications in cytoplasmic actin is still poorly understood. Here we demonstrate that dioxygenase ALKBH4-mediated demethylation of a monomethylated site in actin (K84me1) regulates actin-myosin interaction and actomyosin-dependent processes such as cytokinesis and cell migration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service