Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1250 μL | Check Cart for Availability | $387.00 |
4 x 1250 μL | Check Cart for Availability | $1,400.00 |
About This Item
$387.00
assay
99% (dNTP)
form
solution
usage
sufficient for 125,000 standard reactions (03732720001), sufficient for 31,250 standard reactions (11969056001), sufficient for 6,250 standard reactions (11934554001)
packaging
pkg of 1,250 μL (11969056001 [100 mM]), pkg of 4 × 1,250 μL (03732720001 [4x100 mM]), pkg of 250 μL (11934554001 [100 mM])
manufacturer/tradename
Roche
concentration
<0.9% (dNDP), 100 mM
technique(s)
whole genome amplification: suitable
storage temp.
−20°C
SMILES string
[Na].OC1CC(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N2C=CC(=O)NC2=O
InChI
1S/C9H15N2O14P3.Na.H/c12-5-3-8(11-2-1-7(13)10-9(11)14)23-6(5)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,12H,3-4H2,(H,18,19)(H,20,21)(H,10,13,14)(H2,15,16,17);;
InChI key
FVFWWPFKJNTHIQ-UHFFFAOYSA-N
General description
Application
Physical form
Preparation Note
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description PCR Grade, sodium salt | description ≥85% (HPLC), suitable for ELISA, suitable for hybridization, solution | description PCR Grade, sodium salt | description PCR Grade, sodium salt |
| manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename Roche |
| form solution | form solution | form solution | form liquid |
| assay 99% (dNTP) | assay ≥85% (HPLC) | assay 99% (dNTP) | assay 99% (dNTP) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| concentration <0.9% (dNDP), 100 mM | concentration - | concentration <0.9% (dNDP), 100 mM | concentration <0.9% (dNDP) |
| packaging pkg of 1,250 μL (11969056001 [100 mM]), pkg of 250 μL (11934554001 [100 mM]), pkg of 4 × 1,250 μL (03732720001 [4x100 mM]) | packaging pkg of 25 μL (25 nmol; 1mM) | packaging pkg of 1,250 μL (11969048001 [100 mM]), pkg of 4 × 1,250 μL (03732711001 [4x100 mM]), pkg of 250 μL (11934546001 [100 mM]) | packaging pkg of 4 × 1,250 μL (03622614001 [4x100 mM]), pkg of 4 × 250 μL (11969064001 [4x100 mM]) |
Storage Class
12 - Non Combustible Liquids
flash_point_f
does not flash
flash_point_c
does not flash
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


