Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Check Cart for Availability | $445.00 |
2 x 1 mL | Check Cart for Availability | $758.00 |
About This Item
$445.00
assay
>98%
form
solution
packaging
bottle of 1 mL (11685627001[25 mM]), bottle of 2 × 1 mL (11759051001[25 mM])
manufacturer/tradename
Roche
shipped in
wet ice
storage temp.
2-8°C
SMILES string
Cl[C@]1(C2)C[C@H]3C4(OOC4(C5=CC(OP([O-])([O-])=O)=C(Cl)C=C5)OC)[C@@H]2C[C@@H](C1)C3
InChI
1S/C18H21Cl2O7P.2Na/c1-24-18(14-6-13(2-3-15(14)19)25-28(21,22)23)17(26-27-18)11-4-10-5-12(17)9-16(20,7-10)8-11;;/h2-3,6,10-12H,4-5,7-9H2,1H3,(H2,21,22,23);;/q;2*+1/p-2/t10-,11-,12+,16-,17?,18?;;
InChI key
UKWLRLAKGMZXJC-VCCHNUGISA-L
General description
25 mM solution (12.38 mg/ml), 100x concentration
Application
Features and Benefits
- Save time with a fast, ultra-sensitive substrate.
- Easily strip and reprobe membranes.
- Take multiple exposures of up to two days for each experiment.
- Reuse the substrate by filtering and storing in sodium azide.
Preparation Note
Maleic Acid Buffer: 15 to 25 °C, stable
Detection Buffer: 15 to 25 °C, stable
Blocking Solution: always prepare freshly
Antibody Solution: 2 to 8 °C
Analysis Note
Other Notes
- First exposure to X-ray film should be 5-15 minutes. Examine film and then determine other exposure times based on initial result.
- No preincubation step required.
- Apply substrate quickly to avoid gray shadows from substrate drops and drying of membrane (leads to uneven, high background).
- Do not use plastic wrap to cover blot; use hybridization bags, acetate sheet protectors, or two sheets of transparent film.
- Exposure times are shorter than with CSPD.
- Do not use nitrocellulose membranes.
- Repeat exposures can be made up to two days after the addition of substrate.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| assay >98% | assay ≥75% (HPLC) | assay >98% (HPLC) | assay ≥98% (HPLC) |
| form solution | form liquid | form solution | form powder |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
| packaging bottle of 1 mL (11685627001[25 mM]), bottle of 2 × 1 mL (11759051001[25 mM]) | packaging - | packaging pkg of 1 mL (11655884001[25mM]), pkg of 2 × 50 mL (11755633001[0.25mM] ready-to-use) | packaging - |
| manufacturer/tradename Roche | manufacturer/tradename - | manufacturer/tradename Roche | manufacturer/tradename - |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
No data available
flash_point_c
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![CDP-Star® Disodium 2-chloro-5-(4-methoxyspiro {1,2-dioxetane-3,2′-(5′-chloro)tricyclo[3.3.1.13,7 ]decan}-4-yl)-1-phenyl phosphate](/deepweb/assets/sigmaaldrich/product/structures/224/846/d08963f5-c8ba-42e6-b4f7-9ee7fd76d809/640/d08963f5-c8ba-42e6-b4f7-9ee7fd76d809.png)
![CSPD ready-to-use Disodium 3-(4-methoxyspiro {1,2-dioxetane-3,2′-(5′-chloro)tricyclo [3.3.1.13,7]decan}-4-yl)phenyl phosphate](/deepweb/assets/sigmaaldrich/product/images/352/091/ef743cea-ccd8-44f1-8f3b-dec5a1e4f5d1/640/ef743cea-ccd8-44f1-8f3b-dec5a1e4f5d1.jpg)
