Skip to Content
Merck
All Photos(1)

Key Documents

DCTP-RO

Roche

dCTP

99% (dNTP), <0.9% (dNDP), solution, 100 mM

Synonym(s):

2′-Deoxycytidine 5′-triphosphate disodium salt, dCTP-Na2

Sign Into View Organizational & Contract Pricing

Select a Size

250 μL
$87.90

$87.90


Estimated to ship onApril 08, 2025



Select a Size

Change View
250 μL
$87.90

About This Item

Empirical Formula (Hill Notation):
C9H14N3Na2O13P3
CAS Number:
Molecular Weight:
511.12
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

$87.90


Estimated to ship onApril 08, 2025


description

PCR Grade, sodium salt

Quality Level

Assay

99% (dNTP)

form

solution

usage

sufficient for 125,000 standard reactions (03732690001)
sufficient for 31,250 standard reactions (11969021001)
sufficient for 6,250 standard reactions (11934520001)

packaging

pkg of 1,250 μL (11969021001 [100 mM])
pkg of 4 × 1,250 μL (03732690001 [4x100 mM])
pkg of 250 μL (11934520001 [100 mM])

manufacturer/tradename

Roche

concentration

<0.9% (dNDP)
100 mM

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O2

InChI

1S/C9H16N3O13P3.2Na/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(23-8)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,13H,3-4H2,(H,18,19)(H,20,21)(H2,10,11,14)(H2,15,16,17);;/q;2*+1/p-2/t5-,6+,8+;;/m0../s1

InChI key

ABWVCNMFYVEBIB-CDNBRZBRSA-L

Looking for similar products? Visit Product Comparison Guide

General description

dCTP, PCR Grade, is a 100 mM clear colorless solution of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).

Application

2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.

Quality

Function tested in RT-PCR. Tested for absence of contaminating deoxyribonucleases, ribonucleases and nicking activities according to the current Quality Control procedures.

Physical form

solution of dCTP in water (pH 8.3).

Preparation Note

Working solution: We recommend to prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).

Other Notes

For general laboratory use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahir Karababa et al.
Antimicrobial agents and chemotherapy, 48(8), 3064-3079 (2004-07-27)
Azole resistance in Candida albicans can be due to upregulation of multidrug transporters belonging to ABC (ATP-binding cassette) transporters (CDR1 and CDR2) or major facilitators (CaMDR1). Upregulation of these genes can also be achieved by exposure to fluphenazine, resulting in
Alix T Coste et al.
Eukaryotic cell, 3(6), 1639-1652 (2004-12-14)
The ABC transporter genes CDR1 and CDR2 can be upregulated in Candida albicans developing resistance to azoles or can be upregulated by exposing cells transiently to drugs such as fluphenazine. The cis-acting drug-responsive element (DRE) present in the promoters of
Xiangang Zong et al.
Pflugers Archiv : European journal of physiology, 458(5), 891-899 (2009-06-27)
Second messenger-induced Ca(2+)-release from intracellular stores plays a key role in a multitude of physiological processes. In addition to 1,4,5-inositol trisphosphate (IP(3)), Ca(2+), and cyclic ADP ribose (cADPR) that trigger Ca(2+)-release from the endoplasmatic reticulum (ER), nicotinic acid adenine dinucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service