Skip to Content
Merck
All Photos(4)

Documents

2GFRMKB

Roche

KAPA2G Fast ReadyMix

suitable for PCR, 2 ×, with dye

Synonym(s):

PCR, mastermix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 100 reactions
sufficient for 2000 reactions
sufficient for 500 reactions

shelf life

≤18 mo.

feature

Fast PCR
dNTPs included
hotstart: no

packaging

kit of 1.25 mL (100 x 25 μL rxn; KK5101)
kit of 25 mL (KK5103)
kit of 6.25 mL (500 x 25 μL rxn; KK5102)

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

General description

KAPA2G Fast DNA Polymerase is a second-generation (2G) enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq polymerase. In addition to speed, KAPA2G Fast provides higher yields and sensitivity than competitor enzymes across a broad range of targets.
KAPA2G Fast ReadyMix PCR Kits are designed for fast PCR, in which total reaction times are 20–70% shorter than those of conventional PCR assays performed with wild-type Taq DNA polymerase. This can be achieved without sacrificing reaction performance, and does not require specialized PCR consumables or thermocyclers.

Application

KAPA2G Fast ReadyMix has been used in
  • Nested PCR (nPCR)
  • Routine PCR
  • Fast PCR
  • Genotyping

Biochem/physiol Actions

DNA fragments generated with KAPA2G Fast DNA Polymerase may be used for routine downstream analyses or applications, including restriction enzyme digestion, cloning and sequencing. The fidelity of KAPA2G Fast is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides. Like wild-type Taq, KAPA2G Fast has 5→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity.

Features and Benefits

Reduce cycling times up to 75%:
  • Extension times as low as 1 sec per kilobase
  • Broad coverage of both AT- and GC-rich targets

High speed, higher performance:
  • Single copy sensitivity from complex targets
  • Increased yields

Quick Notes:
  • KAPA2G Fast ReadyMix PCR Kits contain the engineered KAPA2G Fast DNA Polymerase, developed for fast PCR.
  • Use 1 sec extension time for amplicons <1 kb and 15 sec/kb for longer amplicons, and save 20–70% of total reaction time.
  • No need for specialized instrumentation or PCR consumables.
  • Optimized buffer system offers improved yields, specificity and sensitivity, facilitating efficient primerannealing across a wide range of primer lengths, GC contents, and melting temperatures.
  • For high reaction efficiency, do not exceed 25 μL reaction volumes.
  • KAPA2G Fast ReadyMix contains 1.5 mM MgCl2 at 1X.KAPA2G Fast ReadyMix includes two inert tracking dyes to allow direct loading onto agarose gels.
  • Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Fast DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230Assay). KAPA2G Fast ReadyMix is subjected to stringent quality control tests, is free of contaminating exo- andendonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that the ReadyMix hasbeen handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally )at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°Cis not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradationwhen contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA2G Fast HotStart® DNA Polymerase 0.5 U per 25 µL reaction

  • MgCl2 1.5mM at 1X

  • Stabilizers

  • dNTP Mix 0.2 mM of each dNTP at 1X

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative immunological studies between tissue culture and egg adapted duck plague vaccines.
Farouk H, et al.
Report and opinion, 4, 15-38 (2012)
José Vicente Gomes-Filho et al.
RNA biology, 12(5), 490-500 (2015-03-26)
The existence of sense overlapping transcripts that share regulatory and coding information in the same genomic sequence shows an additional level of prokaryotic gene expression complexity. Here we report the discovery of ncRNAs associated with IS1341-type transposase (tnpB) genes, at
Comparative Diagnosis Utilizing Molecular and Serological Techniques of Theileria equi Infection in Distinct Equine Populations in Egypt.
Olfat A M, et al.
International Journal of ChemTech Research, 9(6), 185-197 (2016)
Ivica Petrovic et al.
Frontiers in endocrinology, 11, 30-30 (2020-03-03)
Aims/Hypothesis: Galectin 3 appears to play a proinflammatory role in several inflammatory and autoimmune diseases. Also, there is evidence that galectin 3 plays a role in both type-1 and type-2 diabetes. During obesity, hematopoietic cell-derived galectin 3 induces insulin resistance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service