Skip to Content
Merck
All Photos(1)

Documents

11888412001

Roche

PCR Nucleotide MixPLUS

solution, Roche, pkg of 2 × 100 μL (10 mM each), suitable for RT-PCR

Synonym(s):

PCR | Nucleotide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

form

solution

Quality Level

usage

sufficient for 500 reactions

packaging

pkg of 2 × 100 μL (10 mM each)

manufacturer/tradename

Roche

technique(s)

RT-PCR: suitable

color

colorless

solubility

water: miscible

storage temp.

−20°C

General description

PCR Nucleotide MixPLUS is a clear, colorless solution of the sodium salts of dATP, dCTP, dGTP, each at a concentration of 10 mM, and dUTP at a concentration of 30 mM in Water, PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.
The incorporation of dUTP in place of dTTP allows the degradation of contaminating PCR products from former reactions with Uracil-DNA Glycosylase (UNG) to prevent carryover contamination from previous amplifications.

Application

  • This ready-to-use nucleotide mix is a premixed solution of the sodium salts of dATP, dGTP, and dCTP, each at a concentration of 10 mM, and dUTP at a concentration of 30 mM in water. This mix is optimized for use in all types of amplification reactions: PCR
  • RT-PCR
  • Prevention of carryover contamination
To allow decontamination of PCR or RT-PCR, dUTP in place of dTTP is incorporated into the PCR product. Subsequent reactions may then be treated with Uracil-DNA Glycosylase (UNG). Avoiding the need of re-opening the reaction vial, the vials are incubated at +20 °C, resulting in the degradation of potentially contaminating uracil-containing amplification products. During this step, template DNA and RNA remain unaffected, since normal DNA does not contain uracil, and RNA does not serve as a substrate for UNG. Before starting the actual thermocycling program, UNG is inactivated by incubation at +95 °C. Uracil -DNA Glycosylase, heat-labile is particularily useful, as it is fully inactivated already after incubation at +95 °C for 2 minutes. The natural enzyme from E. coli requires incubating the reaction mixture for 10 minutes at +95 °C. The shorter heat treatment substantially reduces the risk for loosing the template nucleic acid, which typically is present at low concentrations only. This is of particular importance, when performing RT-PCR. We therefore recommend the use of Uracil-DNA Glycosylase.
It has been used in LightCycler PCR assay to identify B. pertussis and B.parapertussis in nasopharyngeal swabs. It has also been used in quantitative PCR (qPCR) assay.

Features and Benefits

The PCR Nucleotide MixPLUS can be added directly to amplification reactions.PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. They can be used to amplify even low amounts of template RNA and DNA.
Improve yield and performance
  • Choose a convenient ready-to-use mix of all 4 nucleotides.
  • Achieve the highest PCR and RT-PCR sensitivity.
  • Insist on highest purity (>99%) nucleotides

Packaging

1 set containing 4 ready-to-use mixes

Quality

Function tested in PCR to ensure specific DNA amplification. Decontamination with UNG is verified. No RNases or DNases are detectable according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • dATP, PCR Grade, sodium salt 10 mM

  • dCTP, PCR Grade, sodium salt 10 mM

  • dGTP, PCR Grade, sodium salt 10 mM

  • dUTP, PCR Grade, sodium salt 10 mM

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynne M Sloan et al.
Journal of clinical microbiology, 40(1), 96-100 (2002-01-05)
A rapid real-time multiplex PCR assay for detecting and differentiating Bordetella pertussis and Bordetella parapertussis in nasopharyngeal swabs was developed. This assay (LC-PCR-IS) targets the insertion sequences IS481 and IS1001 of B. pertussis and B. parapertussis, respectively, and is performed
Miki Kasai et al.
Journal of clinical microbiology, 46(11), 3690-3702 (2008-10-11)
We developed two real-time quantitative PCR (qPCR) assays, targeting the 28S rRNA gene, for the diagnosis of zygomycosis caused by the most common, clinically significant Zygomycetes. The amplicons of the first qPCR assay (qPCR-1) from Rhizopus, Mucor, and Rhizomucor species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service