Skip to Content
Merck

Skip To

08039852001

Roche

Carboxypeptidase B, recombinant

solution, ~150 units/mg protein (at +25°C with hippuryl-L-arginine as the substrate), optimum pH 7.0-9.0

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

EC Number:
NACRES:
NA.21
UNSPSC Code:
12352204
Specific activity:
~150 units/mg protein (at +25°C with hippuryl-L-arginine as the substrate)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

specific activity

~150 units/mg protein (at +25°C with hippuryl-L-arginine as the substrate)

shelf life

A decrease in activity of approx. 10% may occur within 6 months.

mol wt

34.6 kDa

packaging

pkg of 1 mL (2.8 mg)

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

optimum pH

7.0-9.0

storage temp.

−20°C

Application

Peptidyl-L-lysine (-L-arginine) hydrolase
Use Carboxypeptidase B for the sequence analysis of proteins by successive cleavage of basic amino acids from the C-terminus of proteins.

Biochem/physiol Actions

Carboxypeptidase B is a Zn2+-metalloprotease, catalyzing the hydrolysis of the basic amino acids L-Lys and L-Arg from the C-terminal position in polypeptides.

Analysis Note

Contaminants: Contaminants expressed as percentage of carboxypeptidase B activity.
Carboxypeptidase A ≤2%
Trypsin (with Chymotrypsin as substrate) ≤0.005 U/mg

Other Notes

For life science research use only. Not for use in diagnostic precedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP330213173RSTREPRO
description

solution, ~150 units/mg protein (at +25°C with hippuryl-L-arginine as the substrate), optimum pH 7.0-9.0

description

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

description

-

description

liquid, ≥17 units/mg protein, pH range 5.0-9.0

specific activity

~150 units/mg protein (at +25°C with hippuryl-L-arginine as the substrate)

specific activity

-

specific activity

-

specific activity

≥17 units/mg protein

form

solution

form

lyophilized

form

liquid

form

liquid

shelf life

A decrease in activity of approx. 10% may occur within 6 months.

shelf life

-

shelf life

-

shelf life

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

-10 to -25°C

storage condition

avoid repeated freeze/thaw cycles

storage condition

-

storage condition

-

storage condition

-

mol wt

34.6 kDa

mol wt

35.1 kDa

mol wt

Mw ~47 kDa

mol wt

~52 kDa


Storage Class

12 - Non Combustible Liquids

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library