Key Documents
XX4304704
Millipore
Hose adapter, 1/4 in. NPTF to 5/16 in. ID, polypropylene
Accessories for filter holders for sample preparation.
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Pricing and availability is not currently available.
material
polypropylene adapter
Quality Level
I.D.
6.3 mm (0.25 in.)
shipped in
ambient
General description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.
Tri-Clamp (TC) fittings are referred to by convention as 1 1/2", 2", etc. 1 1/2" TC fittings connect to 1" or 1 1/2" O.D. tube sizes. 2" TC fittings connect to 2" O.D. tube size only.