Skip to Content
Merck
All Photos(2)

Key Documents

VCWP01300

Millipore

MCE Membrane Filter, 0.1 μm Pore Size

greener alternative

MF-Millipore, filter diam. 13 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 0.1 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

sustainability

Greener Alternative Product

parameter

0.4 L/min-cm2 air flow rate
1.5 mL/min-cm2 water flow rate
75 °C max. temp.

diam.

13 mm

filter diam.

13 mm

thickness

100 μm

gravimetric extractables

1.5%

refractive index

n/D 1.5

matrix

MF-Millipore

pore size

0.1 μm pore size
74 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥14.1 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: VCWP
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

Microdialysis of DNA and proteins

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service