Skip to Content
Merck
All Photos(4)

Key Documents

SQPAKGARM1

Milli-Q

SQPAK Gard Pack

greener alternative

Removes free chlorine, colloids, particulates from tap water feeding Milli-Q® SP 240 prefilter module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NB.35
Pricing and availability is not currently available.

Quality Level

packaging

pkg of 1 unit

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

compatibility

for use with Milli-Q® SQ 2Series

greener alternative category

General description

To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The SQPAK Gard pretreatment pack protects the reverse osmosis (RO) membrane of the Milli-Q® SQ 2Series production station by efficiently removing water contaminants such as free chlorine, colloids, and particulates from potable water.
Purification technologies include activated carbon to remove free chlorine and a depth filter to remove particulates and colloids.

For use on the Milli-Q® SP 240 Prefilter module to protect the RO membrane of Milli-Q® SR 240/240L/240XL and SQ 240C/240CV systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Features and Benefits

  • Improves Milli-Q® SQ 2Series system performance and extends system lifetime 
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
  • Easy to replace thanks to Twist & Lock mechanism
  • Prefilter label and purple color is repeated on the interface of the Milli-Q® SP 240 Prefilter module to inform about its water purification purpose and guide replacement 

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.

Related Content

Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service