Skip to Content
Merck
All Photos(2)

Key Documents

SMEP0470W

Millipore

MCE Membrane Filter, 5.0 μm Pore Size

MF-Millipore, filter diam. 47 mm, hydrophilic, w/ 6 mm hydrophobic edge

Synonym(s):

MF-Millipore Membrane Filter, Hydrophobic Edge, 5 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

32 L/min-cm2 air flow rate
580 mL/min-cm2 water flow rate
75 °C max. temp.

border W

6 mm , hydrophobic edge

filter diam.

47 mm

thickness

135 μm

matrix

MF-Millipore

pore size

5.0 μm pore size
84 % porosity

bubble point

≥0.42 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: SMEP
Hydrophobic edge membranes are standard MF-Type filters with edges rendered hydrophobic to 3 or 6 mm widths. In antibiotic and bacteriostatic drug sterility testing by membrane filter method, the 6 mm wide non-wetting edge prevents drug intrusion under holder sealing rim.

Applications:
- Microbiological and sterility testing of antibiotics and bacteriostatic drugs
- Filling machine operations
-Available in sterile or autoclave-ready packs

Application

47 mm diameter, mixed cellulose esters (MCE) membrane w/ 6 mm hydrophobic edge, white, 100 discs

Packaging

Blue box with 4 x 25 filters

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service